Skip to Content
Merck
All Photos(1)

Key Documents

10988537001

Roche

7-Deaza-2′-deoxy-guanosine-5′-triphosphate

95% (7-Deaza-dGTP, HPLC), ≤4% (7-Deaza-dGDP, HPLC), solution, 10 mM

Synonym(s):

7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt, −N7-dGTP, 7-Deaza-dGTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N4O13P3
CAS Number:
Molecular Weight:
506.19
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

Quality Level

form

solution

composition

7-Deaza-dGDP, ≤4% HPLC
7-Deaza-dGTP, 95% HPLC

packaging

pkg of 200 μL (10 mM, 2 μmol)

manufacturer/tradename

Roche

concentration

10 mM

storage temp.

−20°C

SMILES string

NC1=NC(=O)c2ccn(C3CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O3)c2N1

InChI

1S/C11H17N4O13P3/c12-11-13-9-5(10(17)14-11)1-2-15(9)8-3-6(16)7(26-8)4-25-30(21,22)28-31(23,24)27-29(18,19)20/h1-2,6-8,16H,3-4H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,12,13,14,17)

InChI key

DLLXAZJTLIUPAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-deaza-2′-deoxy-guanosine-5′-triphosphate is a nucleotide analog that is a candidate substrate for processive telomerase. The use of this analog improves PCR (polymerase chain reaction) yields. This compound also makes sequencing of GC rich sequences easier, especially if template amount is low and/or DNA quality is not good.
The product contains 7-Deaza-2′-deoxy-guanosine-5′-triphosphate as a 10 mM solution of the lithium salt.

Application

7-Deaza-dGTP is a substrate for most DNA polymerases, including Taq DNA polymerase.
7-Deaza-dGTP is used in the dideoxy-chain termination sequencing methods, in place of dGTP to overcome compression problems in gel electrophoresis when sequencing GC-rich stretches of DNA.
Partial substitution of 7-Deaza-dGTP for dGTP in PCRs can improve the product yield when the template is GC-rich (i.e., has extensive secondary structures).

Preparation Note

Working solution: Dilute 7-deaza-dGTP into a solution containing 50 mM Tris, pH 7.0, and use at exactly the same concentration as you would use dGTP.

Analysis Note

Absorption: 25 A260 units of product correspond to 1 mg.
Absorbance determination: A250/A260, A280/A260, A290/A260

Other Notes

When PCR products contain 7-Deaza dGTP, they do not stain well in ethidium bromide, presumably because stacking of adjacent bases in the products is diminished.
PCR amplification efficiency is generally not affected by 7-Deaza-dGTP.
For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Johnson et al.
Molecular human reproduction, 17(12), 721-726 (2011-08-13)
Intact ribosomal RNAs (rRNAs) comprise the majority of somatic transcripts, yet appear conspicuously absent in spermatozoa, perhaps reflecting cytoplasmic expulsion during spermatogenesis. To discern their fate, total RNA retained in mature spermatozoa from three fertile donors was characterized by Next
A Jung et al.
Molecular pathology : MP, 55(1), 55-57 (2002-02-12)
CpG islands are GC rich sequences that are found in the promoters of many genes in higher eukaryotes. They contain a high frequency of CG dinucleotides, which are substrates for DNA methylases. Methylation leads to transcriptional silencing of promoters. Owing
Tomonori Yabuta et al.
International journal of cancer, 101(5), 434-441 (2002-09-07)
To identify germline E-cadherin mutations responsible for the predisposition to diffuse gastric cancer (DGC) among the Japanese, we screened 17 patients with familial aggregation of gastric cancer by sequencing analysis. All the patients were diagnosed with DGC and had at
Apurva Agrawal et al.
Recent patents on anti-cancer drug discovery, 7(1), 102-117 (2011-08-23)
Telomerase, a specialised RNA-directed DNA polymerase extends and stabilises the telomeres at the ends of the eukaryotic chromosomes. The progressive loss of telomeres results in limited number of cell divisions and has been linked to the mechanism of human cellular
Suzanne Lesage et al.
Brain : a journal of neurology, 136(Pt 2), 385-391 (2013-02-16)
The recently identified C9orf72 gene accounts for a large proportion of amyotrophic lateral sclerosis and frontotemporal lobar degenerations. As several forms of these disorders are associated with parkinsonism, we hypothesized that some patients with Parkinson's disease or other forms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service