Skip to Content
Merck
All Photos(2)

Documents

D8783

Sigma-Aldrich

7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt

10 mM in H2O

Synonym(s):

−N7-dGTP, 7-Deaza-dGTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N4O13P3
CAS Number:
Molecular Weight:
506.19
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52

biological source

Porcine muscle
rabbit muscle

Assay

≥95% (HPLC)

form

liquid

concentration

10 mM in H2O

color

colorless

shipped in

dry ice

storage temp.

−20°C

SMILES string

NC1=NC(=O)c2ccn(C3CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O3)c2N1

InChI

1S/C11H17N4O13P3/c12-11-13-9-5(10(17)14-11)1-2-15(9)8-3-6(16)7(26-8)4-25-30(21,22)28-31(23,24)27-29(18,19)20/h1-2,6-8,16H,3-4H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,12,13,14,17)

InChI key

DLLXAZJTLIUPAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt has been used as a lysis agent to evaluate its efficiency on direct cell lysis, RNA stability, and compatibility with downstream reverse transcription quantitative real-time PCR during the analyzes of samples with small numbers of cells.

Biochem/physiol Actions

7-Deaza-2′-deoxyguanosine 5′-triphosphate (7-deaza-dGTP) competes with natural substrate dGTP and blocks the telomerase activity. This nucleotide once incorporated into the telomere alters the secondary structure of the telomere making it difficult for telomerase to identify it for further telomere synthesis. This dGTP analog pairs weakly with conventional bases to minimize DNA secondary structures, while being a good substrate for DNA polymerases. 7-deaza-dGTP in combination with dimethyl sulfoxide (DMSO) and betaine enhances the polymerase chain reaction (PCR) amplification of GC-rich DNA sequences.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuo Hata et al.
The Journal of molecular diagnostics : JMD, 20(1), 46-55 (2017-12-13)
Telomere end-to-end fusions are an important source of chromosomal instability that arise in cells with critically shortened telomeres. We developed a nested real-time quantitative PCR method for telomere fusion detection in pancreatic ductal adenocarcinomas, intraductal papillary mucinous neoplasms (IPMNs), and
Andreas Blecha et al.
Microbial cell factories, 4, 28-28 (2005-10-06)
Native as well as recombinant bacterial cell surface layer (S-layer) protein of Geobacillus (G.) stearothermophilus ATCC 12980 assembles to supramolecular structures with an oblique symmetry. Upon expression in E. coli, S-layer self assembly products are formed in the cytosol. We
David Svec et al.
Frontiers in oncology, 3, 274-274 (2013-11-14)
The interest to analyze single and few cell samples is rapidly increasing. Numerous extraction protocols to purify nucleic acids are available, but most of them compromise severely on yield to remove contaminants and are therefore not suitable for the analysis
Paulina H Wanrooij et al.
Nucleic acids research, 40(20), 10334-10344 (2012-09-12)
In human mitochondria the transcription machinery generates the RNA primers needed for initiation of DNA replication. A critical feature of the leading-strand origin of mitochondrial DNA replication is a CG-rich element denoted conserved sequence block II (CSB II). During transcription
Emmanuel F Mongodin et al.
PLoS genetics, 2(12), e214-e214 (2006-12-30)
Arthrobacter sp. strains are among the most frequently isolated, indigenous, aerobic bacterial genera found in soils. Member of the genus are metabolically and ecologically diverse and have the ability to survive in environmentally harsh conditions for extended periods of time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service