This product is a 1 mg/mL solution. The enzymatic activity of each component is as follows:
Dispase Activity: 1.0 units per milligram
Collagenase Activity: ≥12,500 units per vial
Caseinase Activity: ≥2000 caseinase units/mg
Clostripain Activity: ≤5.0 clostripain units/mg
Tryptic Activity: ≤0.5 tryptic units/mg
SCM133
PluriSTEM® Dispase-II Solution
Dispase-II has proven to be a rapid and effective, yet gentle, agent for separating many tissues and cells grown in vitro.
Synonym(s):
Dispase, Dispase 2
About This Item
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
PluriSTEM®
concentration
1 mg/mL in sterile DMEM/F-12
technique(s)
cell culture | mammalian: suitable
cell culture | stem cell: suitable
shipped in
dry ice
General description
PluriSTEM Dispase-II has been validated to work alongside PluriSTEM Human ES/iPS Medium (SCM130) for the culture and passage of human embryonic and induced pluripotent stem cells.
Dispase-II enzyme is produced in Bacillus polymyxa.
Application
1. Coat 6-well plates with 1:20 Matrigel coating (1.5 ml per well). Incubate at 2-8ºC overnight or coat at RT for at least 30 minutes to 1 hour before use.
2. Remove matrigel coating. Add 2 mL of Complete Media per well.
3. Remove areas of differentiation within culture.
4. Aspirate media.
5. Wash once with 2 mL PBS.
6. Add 1.5 mL Dispase II per well. Incubate for 37ºC for 7 minutes.
7. Wash 2X with 2 mL PBS, w/o Mg and Ca.
8. Add 2 mL Complete Media to each well and use cell scraper to detach colonies.
9. Collect scrapped cells in 15 ml conical tube.
10. Spin 800 rpm for 5 minutes
11. Resuspend in appropriate volume of complete media. Typical splitting ratio is 1:5 – 1:6 depending upon cell density. Cells should be ready for passaging in 5-6 days time using PluriSTEM Human ES/iPS Medium.
Stem Cell Research
Enzymes & Biochemistry
Quality
Physical form
Dispase-II enzyme is produced in Bacillus polymyxa.
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the activity of this enzime?
1 answer-
Helpful?
-
-
What is the activity of the dispase product SCM133, and is there a specific range it should fall within in units/mL?
1 answer-
The activity of the material is 1.0 unit per milligram (u/mg).
Helpful?
-
-
Please let me know U/ml of this (https://www.sigmaaldrich.com/CZ/en/product/mm/scm133) solution. Thank you
1 answer-
This product is a 1 mg/mL solution of Dispase II. The enzyme activity is lot specific, however the activity will be ?0.8 U/mg. This activity is established using casein as a substrate at 37°C and pH 7.5. Please see the link below to review the product data sheet:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/727/543/scm133.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service