SCC009
ReNcell CX Human Neural Progenitor Cell Line & Culture Media Kit
ReNcell CX Kit contains human neural progenitor cells, optimized maintenance media & freezing media for culture of human neural stem cells.
Synonym(s):
Human NSC Line, Human Neural Stem Cell Line, Human Neural Stem Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
manufacturer/tradename
Chemicon®
ReNcell
technique(s)
cell culture | stem cell: suitable
input
sample type neural stem cell(s)
shipped in
liquid nitrogen
General description
ReNcell CX Kit contains human neural progenitor cells, optimized maintenance media and freezing media for culture of human neural stem cells. ReNcell CX Immortalized Cells (SCC007): ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells. ReNcell CX was derived from the cortical region of human fetal brain. Immortalized by retroviral transduction with the c-myc oncogene, this cell line grows rapidly as a monolayer on laminin with a doubling time of 20-30 hours. Karyotype analyses indicate that the ReNcell CX retains a normal diploid karyotype in culture even after prolonged passage (>45 passages). ReNcell CX was developed by the ReNeuron Group plc, a biotech company that specializes in using human somatic stem cells for therapeutics. ReNcell CX may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ReNcell CX cells have been obtained in a legal and ethical manner, compliant with current local informed consent procedures. ReNcell NSC Maintenance Medium (SCM005): ReNcell Neural Stem Cell (NSC) Maintenance Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells. When used in conjunction with FGF and EGF, the maintenance medium will allow for the proliferation of ReNcell immortalized VM and CX neural stem cells. Withdrawal of the growth factors from ReNcell NSC Maintenance Medium will result in the spontaneous differentiation of ReNcell immortalized neural progenitor cells. ReNcell NSC Freezing Medium (SCM007): ReNcell NSC Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX (CHEMICON Catalog No. SCC007) and VM (CHEMICON Catalog No. SCC008) cultured in serum-free conditions with ReNcell NSC Maintenance Medium (CHEMICON Catalog No. SCM005). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.
Cell Line Description
Neural Stem Cells
Application
Research Category
Stem Cell Research
Stem Cell Research
Components
ReNcell Neural Stem Cell Freezing Medium (Cat.# SCM007)
ReNcell NSC Maintenance Media (Cat.# SCM005)
ReNcell CX Immortalized Cell Line (Cat.# SCC007)
ReNcell NSC Maintenance Media (Cat.# SCM005)
ReNcell CX Immortalized Cell Line (Cat.# SCC007)
Quality
Each lot of ReNcell CX cells has been validated for high level of expression of Nestin and Sox 2 and for their self-renewal and multi-lineage differentiation capacities (please refer to datasheet figures). Cells also display normal karyotype as assessed by chromosome spread and tested negative for mycoplasma. Maternal blood has been screened for HIV, HTLV, Hepatitis B and C.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the "Academic Use Agreement" as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Experimental and therapeutic medicine, 23(2), 164-164 (2022-01-25)
Bisphenol A (BPA) is a common industrial chemical widely used to produce various plastics and is known to impair neural stem cells (NSCs). However, the effects of low-dose BPA exposure on the stemness maintenance and differentiation fate of NSCs remain
Differential development of neuronal physiological responsiveness in two human neural stem cell lines.
BMC Neuroscience null
Developmental cell, 46(1), 73-84 (2018-07-06)
The ability to convert human somatic cells efficiently to neurons facilitates the utility of patient-derived neurons for studying neurological disorders. As such, ectopic expression of neuronal microRNAs (miRNAs), miR-9/9∗ and miR-124 (miR-9/9∗-124) in adult human fibroblasts has been found to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service