Skip to Content
Merck
All Photos(2)

Key Documents

MABN140

Sigma-Aldrich

Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4

clone 27-4, from rabbit

Synonym(s):

Rabbit Anti-NeuN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

27-4, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBFOX3(146713)
mouse ... Rbfox3(52897)

General description

NeuN is a RNA-binding protein that belongs to the evolutionarily conserved Fox-1 family. It is mainly involved RNA splicing. It is found exclusively in the nuclei of neuronal cells, but is also present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although. Some neurons fail to recognize NeuN at all ages in certain cells. Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron. Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found.

Immunogen

Recombinant protein corresponding to mouse NeuN.

Application

Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4 detects level of NeuN (rabbit monoclonal) & has been published & validated for use in ICC, Western Blotting & IHC.
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected NeuN in rat E18 cortex and hippocampus primary cultures.

Immunohistochemistry Analysis: A 1:400 dilution of this antibody detected NeuN in mouse cerebellum and hippocampus tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse brain nuclear extract.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected NeuN in 10 µg of mouse brain nuclear extract.

Target description

~46/48 kDa observed

Physical form

Format: Purified
Protein A purified
Purified rabbit monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain nuclear extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aran Groves et al.
eNeuro, 5(5) (2018-09-27)
Astrocytes have prominent roles in central nervous system (CNS) function and disease, with subpopulations defined primarily by morphologies and molecular markers often determined in cell culture. Here, we identify an in vivo astrocyte subpopulation termed immediate-early astrocytes (ieAstrocytes) that is
Rahul Agrawal et al.
JCI insight, 4(20) (2019-10-18)
It is proposed that the impaired sympathoadrenal response to hypoglycemia induced by recurrent insulin-induced hypoglycemia (RH) is an adaptive phenomenon induced by specific changes in microRNA expression in the ventromedial hypothalamus (VMH). To test this hypothesis, genome-wide microRNAomic profiling of
Michael D Scofield et al.
Biological psychiatry, 78(7), 441-451 (2015-04-12)
Glial cells of the central nervous system directly influence neuronal activity by releasing neuroactive small molecules, including glutamate. Long-lasting cocaine-induced reductions in extracellular glutamate in the nucleus accumbens core (NAcore) affect synaptic plasticity responsible for relapse vulnerability. We transduced NAcore
Cassandra L Schlamp et al.
Molecular vision, 19, 1387-1396 (2013-07-05)
Retinal ganglion cells comprise a percentage of the neurons actually residing in the ganglion cell layer (GCL) of the rodent retina. This estimate is useful to extrapolate ganglion cell loss in models of optic nerve disease, but the values reported
Hongyu Pan et al.
Stem cell reports, 7(4), 707-718 (2016-10-04)
Adult hippocampal neurogenesis was impaired in several Alzheimer's disease models overexpressing mutant human amyloid precursor protein (hAPP). However, the effects of wild-type hAPP on adult neurogenesis and whether the impaired adult hippocampal neurogenesis was caused by amyloid β (Aβ) or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service