Skip to Content
Merck
All Photos(1)

Key Documents

MAB5416

Sigma-Aldrich

Anti-Glutamate Receptor 3 Antibody, clone 3B3

clone 3B3, Chemicon®, from mouse

Synonym(s):

AMPA-selective glutamate receptor 3, Glutamate receptor ionotropic, AMPA 3, glutamate receptor 3, glutamate receptor C, glutamate receptor subunit 3, glutamate receptor, ionotrophic, AMPA 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B3, monoclonal

species reactivity

monkey, mouse, human

species reactivity (predicted by homology)

rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIA3(2892)

General description

AMPA receptors mediate fast synaptic current at most excitatory synapses, with stoichiometry characterized by subtype composition.There are four types of AMPA selective GluR subunits (GluR1, GluR2, GluR3 and GluR4). Tetrameric or pentameric combinations of different subunits contributes to the functional diversity of AMPA receptors. The insertion or removal of GluR1/GluR4 oligomeric channels from postsynaptic membranes appears to be LTP/LTD activity dependent while GluR2/GluR3 oligomers are continuously cycling. Millipore’s anti-GluR2/3 antibody is useful for localizing expression of GluR2/GluR3 receptor assemblies in the PSD.

Specificity

Reacts with Glutamate Receptor 3 (GluR3). No cross reactivity with GluR2.

Immunogen

Fusion protein, N-terminus of GluR3

Application

Anti-Glutamate Receptor 3 Antibody, clone 3B3 is an antibody against Glutamate Receptor 3 for use in WB, IC, IH, IH(P).
Immunohistochemistry: A previous lot of this antibody was used in IH.

Optimal working dilutions must be determined by end user.

Quality

Western Blot Analysis: A previous lot of this antibody was used in WB.

Target description

~ 110 kDa

Physical form

Format: Purified
Liquid in PBS. Contains no preservative.

Analysis Note

Control
Brain

Hippocampal tissue

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Bartolomé-Martín et al.
Journal of cell science, 125(Pt 2), 422-434 (2012-02-15)
Following the exocytosis of neurotransmitter-containing synaptic vesicles, endocytosis is fundamental to re-establishing conditions for synaptic transmission. As there are distinct endocytotic pathways that each differ in their efficiency to generate releasable synaptic vesicles, we used the dye FM1-43 to track
Evolutionarily conserved pattern of AMPA receptor subunit glycosylation in Mammalian frontal cortex.
Tucholski, J; Pinner, AL; Simmons, MS; Meador-Woodruff, JH
Testing null
Xiu Sun et al.
The European journal of neuroscience, 30(4), 539-550 (2009-08-14)
Synaptic scaling has been proposed as a form of plasticity that may contribute to drug addiction but it has not been previously demonstrated in the nucleus accumbens (NAc), a critical region for addiction. Here we demonstrate bidirectional synaptic scaling in
Janusz Tucholski et al.
Schizophrenia research, 146(1-3), 177-183 (2013-03-07)
Numerous studies have demonstrated brain region- and subunit-specific abnormalities in the expression of subunits of the AMPA subtype of glutamate receptors in schizophrenia. In addition, abnormalities in the expression of proteins that regulate the forward trafficking of AMPA receptors through
Maya Yamazaki et al.
The European journal of neuroscience, 31(12), 2204-2220 (2010-06-10)
The alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)-type glutamate receptors require auxiliary subunits termed transmembrane AMPA receptor regulatory proteins (TARPs), which promote receptor trafficking to the cell surface and synapses and modulate channel pharmacology and gating. Of six TARPs, gamma-2 and gamma-7 are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service