Skip to Content
Merck
All Photos(1)

Key Documents

ECM540

Sigma-Aldrich

ECM Cell Adhesion Array Kit, colorimetric

The ECM Cell Adhesion Array kit is useful for assessing specific cell surface integrins, monitoring in vitro cell differentiation, or screening potential cell adhesion promoters/inhibitors.

Synonym(s):

Extracellular matrix array cell adhesion assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

detection method

colorimetric

shipped in

wet ice

General description

Introduction:

Cell adhesion plays a major role in cellular communication and regulation, and is of fundamental importance in the development and maintenance of tissues. Scientists are continually examining the adhesion, migration, and invasion of many diverse cell types on various extracellular matrix (ECM) component proteins. The CHEMICON ECM Cell Adhesion Array Kit (Colorimetric) provides an ECM Array microtiter plate with a colorimetric detection format, allowing for large-scale screening and quantitative comparison of multiple samples/cell lines. The kit is designed as a rapid (1-2 hours), cost effective, efficient method for characterization of cell adhesion.

In addition, Chemicon continues to provide numerous migration, invasion, and adhesion products including:

· Alpha Integrin-Mediated Cell Adhesion Array Kit (Colorimetric) (ECM530)

· Beta Integrin-Mediated Cell Adhesion Array Kit (Colorimetric) (ECM531)

· Alpha/Beta Integrin-Mediated Cell Adhesion Array Combo Kit (Colorimetric) (ECM532)

· Alpha Integrin-Mediated Cell Adhesion Array Kit (Fluorimetric) (ECM533)

· Beta Integrin-Mediated Cell Adhesion Array Kit (Fluorimetric) (ECM534)

· Alpha/Beta Integrin-Mediated Cell Adhesion Array Combo Kit (Fluorimetric) (ECM535)

· ECM Cell Adhesion Array Kit (Fluorimetric) (ECM545)

· QCM 8μm 96-well Chemotaxis Cell Migration Assay (ECM510)

· QCM 5μm 96-well Chemotaxis Cell Migration Assay (ECM512)

· QCM 3μm 96-well Chemotaxis Cell Migration Assay (ECM515)

· QCM 96-well Cell Invasion Assay (ECM555)

· QCM 96-well Collagen-Based Cell Invasion Assay (ECM 556)

· 24-well Insert Cell Migration and Invasion assay systems

· CytoMatrix Cell Adhesion strips (ECM protein coated)

· QuantiMatrix ECM protein ELISA kits



Test Principle:

The CHEMICON ECM Cell Adhesion Array kit utilizes a colorimetric detection format, allowing large-scale screening and quantitative comparison of multiple samples. Each kit contains a 96-well, microtiter plate consisting of 12 X 8-well removable strips. Each well within a strip (7 wells total) is pre-coated with a different ECM protein (see table below) along with one BSA-coated well (negative control). Experimental cells are seeded onto the coated substrates, where adherent cells are captured. Subsequently, unbound cells are washed away, and the adherent cells are fixed and stained. After stain extraction, the relative cell attachment is determined using absorbance readings.



Additional information about the ECM components provided in the kit:

Application

Research Category
Cell Structure
The CHEMICON ECM Cell Adhesion Array kit is useful for assessing specific cell surface integrins, monitoring in vitro cell differentiation, or screening potential cell adhesion promoters/inhibitors.



For Research Use Only. Not for use in diagnostic procedures.

Packaging

96 wells

Components

ECM Array Plate: (PN: 90652) One 96-well plate with 12 strips. Each 8-well strip consists of 7 different human, ECM protein-coated wells (Collagen I, Collagen II, Collagen IV, Fibronectin, Laminin, Tenascin, Vitronectin) and one BSA-coated well (negative control). See the plate layout on Data sheet.

Cell Stain Solution: (Part No. 90144) One bottle - 20 mL.

Extraction Buffer: (Part No. 90145) One bottle - 20 mL.

Assay Buffer: (Part No. 90601) One bottle - 100 mL.

Storage and Stability

The ECM Array Plate can be stored at 2° to 8°C in the foil pouch up to its expiration date. Unused strips may be placed back in the pouch for storage. Ensure that the desiccant remains in the pouch, and that the pouch is securely closed. Keep the remaining kit components at 2° to 8°C.

Precautions:

· Cell stain contains minor amounts of crystal violet, a toxic substance, which may cause cancer and heritable genetic damage. Handle with caution. Toxic by inhalation and if swallowed. Irritating to eyes, respiratory system and skin.

· Extraction buffer contains alcohol. Avoid internal consumption.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srikanth Appikonda et al.
The Journal of biological chemistry, 293(19), 7476-7485 (2018-03-11)
Proteins with domains that recognize and bind post-translational modifications (PTMs) of histones are collectively termed epigenetic readers. Numerous interactions between specific reader protein domains and histone PTMs and their regulatory outcomes have been reported, but little is known about how
Hala Kufaishi et al.
Reproductive sciences (Thousand Oaks, Calif.), 23(7), 931-943 (2016-01-15)
This study tested a hypothesis that primary human vaginal cells derived from tissue of premenopausal women with severe pelvic organ prolapse (POP-HVCs) would display differential functional characteristics as compared to vaginal cells derived from asymptomatic women with normal pelvic floor
Zhao Jingjing et al.
Naunyn-Schmiedeberg's archives of pharmacology, 393(8), 1549-1558 (2020-01-05)
Diffuse large B-cell lymphoma (DLBCL) is the most aggressive non-Hodgkin lymphoma (NHL), accounting for about 31% of the newly diagnosed NHL worldwide. Although approximately 60% of patients who initially received a standard R-CHOP treatment likely have a 3-year event-free survival
Stephen N Greenhalgh et al.
The American journal of pathology, 189(2), 258-271 (2018-11-19)
Recent fate-mapping studies in mice have provided substantial evidence that mature adult hepatocytes are a major source of new hepatocytes after liver injury. In other systems, integrin αvβ8 has a major role in activating transforming growth factor (TGF)-β, a potent
Victoria Florea et al.
PloS one, 8(9), e73146-e73146 (2013-09-17)
The proto-oncogene c-Myc is vital for vascular development and promotes tumor angiogenesis, but the mechanisms by which it controls blood vessel growth remain unclear. In the present work we investigated the effects of c-Myc knockdown in endothelial cell functions essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service