Skip to Content
Merck
All Photos(1)

Key Documents

CBL66

Sigma-Aldrich

Anti-Atrial Natriuretic Peptide Antibody, clone A6

clone A6, Chemicon®, from mouse

Synonym(s):

ANP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

A6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... NPPA(4878)

Specificity

ANP (Whole molecule and residues 4-28)

FUSION PARTNER: NSO myeloma cell line

Immunogen

ANP/BSA conjugate

Application

Competetive radioimmunoassay

(Clinical measurement of circulating ANP levels possibly in conjunction with cyclic GMP)

Liquid phase assay without plasma extraction

Staining of formalin-fixed wax embedded tissue

Optimal working dilutions must be determined by the end user.
Detect Atrial Natriuretic Peptide using this Anti-Atrial Natriuretic Peptide Antibody, clone A6 validated for use in IH(P), RIA.
Research Category
Neuroscience
Research Sub Category
Hormones & Receptors

Physical form

Format: Purified
The monoclonal is presented at a concentration of 1mg in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrastructural and immunocharacterization of undifferentiated myocardial cells in the developing mouse heart.
Zhang, F; Pasumarthi, KB
Journal of Cellular and Molecular Medicine null
E Farina-Lipari et al.
European journal of histochemistry : EJH, 47(2), 133-138 (2003-06-05)
In this work, we showed the presence of atrial natriuretic factor (ANF) in human prostate and compared its localisation in normal and hyperplastic conditions. ANF was localised in epithelial and stromal cells, being increased in hyperplasia, mainly in the stromal
Assessment of embryonic myocardial cell differentiation using a dual fluorescent reporter system.
McMullen, NM; Zhang, F; Pasumarthi, KB
Journal of Cellular and Molecular Medicine null
T V Sukhacheva et al.
Arkhiv patologii, 79(4), 3-12 (2017-08-10)
to analyze the morphological features of the myocardium of the atrial appendages in patients with different forms of atrial fibrillation (AF) and to compare the findings with the clinical parameters of patients. Light and electron microscopies were used to examine
Martijn Wehrens et al.
Cell reports, 39(6), 110809-110809 (2022-05-12)
Hypertrophic cardiomyopathy (HCM) is a genetic heart disease that is characterized by unexplained segmental hypertrophy that is usually most pronounced in the septum. While sarcomeric gene mutations are often the genetic basis for HCM, the mechanistic origin for the heterogeneous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service