Skip to Content
Merck
All Photos(1)

Key Documents

CBL52

Sigma-Aldrich

Anti-Insulin-like Growth Factor-I Antibody, clone M23

clone M23, Chemicon®, from mouse

Synonym(s):

IGF-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

100
300

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

M23, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... IGF1(3479)

Specificity

The basic IGF-1-like forms (pl>9.0) showed substantial cross-reactivity, as did rat and mouse sera. Monoclonal antibody BM23 was capable of abolishing the biological activities of IGF-1 in vitro bioassays for insulin-like, mitogenic and sulphation activities.

AFFINITY CONSTANT: The affinity of monoclonal antibody BM23 for IGF-1 was 1 x 10E10 L/mole and the cross reactivities of IGF-2, MSA and porcine insulin were 1%, 0.05% and 0.0001% respectively

Immunogen

Purified human IGF-1.

Application

Immunohistological staining of tissue sections on frozen or formalin fixed paraffin wax embedded sections

Optimal working dilutions must be determined by the end user.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
This Anti-Insulin-like Growth Factor-I Antibody, clone M23 is validated for use in IH(P) for the detection of Insulin-like Growth Factor-I.

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Aslan et al.
Medical science monitor : international medical journal of experimental and clinical research, 25, 5510-5517 (2019-07-26)
BACKGROUND There is no study in the literature investigating the expression levels of WT1, p53, and IGF-1 in colon polyp subtypes. In this study, we aimed to investigate the expression levels of IGF-1, p53, and WT1 in colon polyp subtypes
E Maiorano et al.
Applied immunohistochemistry & molecular morphology : AIMM, 8(2), 110-119 (2000-08-11)
Insulin-like growth factor 1 (IGF-1) likely is involved in thyrocyte proliferation via autocrine mechanisms, but limited data are available on its in vivo expression in thyroid neoplasms. This prompted us to explore IGF-1 expression at the protein and mRNA levels
An immunoenzymatic method to measure IGF-1 in milk.
A Guidi, L Castigliego, G Iannone, A Armani, D Gianfaldoni
Veterinary Research Communications null
Hakryul Jo et al.
Molecular and cellular biology, 28(13), 4285-4299 (2008-05-07)
Cancer cells in their respective microenvironments must endure various growth-constraining stresses. Under these conditions, the cancer cell-derived factors are thought to modulate the signaling pathways between cell growth and dormancy. Here, we describe a cancer cell-derived regulatory system that modulates
Y-F Huang et al.
British journal of cancer, 99(7), 1096-1102 (2008-09-11)
This study was aimed to identify the expression and the correlation of insulin-like growth factor-1 (IGF-1) system and their prognostic impacts in cervical cancer. Seventy-two patients with early-stage cervical cancer were eligible. We obtained the serum levels of total IGF-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service