Skip to Content
Merck
All Photos(1)

Key Documents

AX0120

Supelco

Acetone

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Dimethyl Ketone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-662-2
grade:
ACS reagent
for extraction
reagent grade
Assay:
≥99.5% (corrected for water content, GC)
technique(s):
UV/Vis spectroscopy: suitable
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
for extraction
reagent grade

Quality Level

Agency

meets USP/NF

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

description

for analytical purposes

Assay

≥99.5% (corrected for water content, GC)

form

liquid

expl. lim.

13.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0003 meq/g Titrable acid
≤0.0006 meq/g Titrable base
≤0.002% Aldehyde (as HCHO)
≤0.05% Isopropyl alcohol
≤0.05% Methanol
≤0.5% Water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

AX0120P-4 is packaged in a poly-coated glass bottle
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Aldehyde (as HCHO): 0.002% max
Assay (GC, corrected for water content): 99.5% min
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Isopropyl alcohol: 0.05% max
Methanol: 0.05% max
Solubility in water: To pass test
Substances reducing permanganate: To pass test
Titrable acid: 0.0003meq/g max
Titrable base: 0.0006meq/g max
Water (H2O): 0.5% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service