Skip to Content
Merck
All Photos(1)

Key Documents

AB745

Sigma-Aldrich

Anti-Collagen Type I (COL1A1) Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Anti-CAFYD, Anti-EDSARTH1, Anti-EDSC, Anti-OI1, Anti-OI2, Anti-OI3, Anti-OI4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Collagen Type I Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin (Semi-purified)

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

Specificity

Recognizes Human collagen type I. Some cross reactivity with other proteins.

Immunogen

Human placental collagen type I

Application

ELISA on human collagen type I: 1:4,000.

Indirect microimmunofluorescent visualization of collagen type I on cryostat sections of human tissue or cultured cells: 1:10-1:40.

Western blot 1:200

Not recommended for use in paraffin treated tissues.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Collagen Type I Antibody is validated for use in ELISA, IH, WB for the detection of Collagen Type I.

Physical form

Ammonium sulfater precipitation followed by DEAE chromatography, and immunodeletion
Format: Semi-Purified
Immunoglobulin fraction, prepared by ammonium sulfate precipitation and chromatography on DEAE-cellulose, of antiSerum cross-absorbed over immobilized human albumin, immunoglobulin and collagen types III, IV and V.

Product is liquid in 0.15M Sodium Chloride, 10mM Sodium Phosphate pH 7.5 with 0.1% Mannitol and 0.1% Dextran as stabilizers. No preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Schieker et al.
Journal of anatomy, 204(2), 133-139 (2004-03-23)
In stem-cell research a major difficulty is caused by the lack of distinctive features that allow the identification of human mesenchymal stem cells (hMSC). Until now, there has been no specific marker and the most common way to identify hMSC
M Fairuz B Jamaluddin et al.
Endocrinology, 159(7), 2656-2669 (2018-05-23)
Uterine leiomyomas (fibroids) are the most common benign tumors that are associated with increased production of extracellular matrix (ECM). Excessive ECM deposition plays a major role in the enlargement and stiffness of these tumors and contributes to clinical symptoms, such
Bo Ma et al.
International journal of biological sciences, 14(4), 437-448 (2018-05-05)
Connective tissue growth factor (CTGF) is a crucial factor that plays a major role in the process of posterior capsule opacification (PCO). However, the effects of CTGF on the proliferation and migration of lens epithelial cells (LECs) and on the
M Navab et al.
The Journal of clinical investigation, 87(5), 1763-1772 (1991-05-01)
Medium from cocultures of human aortic endothelial cells (HAEC) and smooth muscle cells (HASMC) taken from the same donor contained approximately two- to fourfold more macrophage colony-stimulating factor, granulocyte/macrophage colony-stimulating factor, and up to 5.1-fold more transforming growth factor beta
John A Marwick et al.
SLAS discovery : advancing life sciences R & D, 26(9), 1091-1106 (2021-06-04)
Lung imaging and autopsy reports among COVID-19 patients show elevated lung scarring (fibrosis). Early data from COVID-19 patients as well as previous studies from severe acute respiratory syndrome, Middle East respiratory syndrome, and other respiratory disorders show that the extent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service