Skip to Content
Merck
All Photos(1)

Documents

234167

Sigma-Aldrich

Anti-Collagen Type I Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Collagen I Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~115 kDa (doublet) type I collagen protein.
Recognizes type I collagen. Exhibits slight cross-reactivity with type III collagen.
This Anti-Collagen Type I Rabbit pAb is validated for use in Frozen Sections, Immunoblotting, Immunofluorescence, Immunoprecipitation for the detection of Collagen Type I.

Immunogen

Fetal Mouse Skin
purified, fetal mouse skin collagen type I

Application

Frozen Sections (1:30-1:60, see comments)

Immunoblotting (1:200-1:500)

Immunofluorescence (1:20-1:40)

Immunoprecipitation (1:20-1:60)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Romanos, G.E., et al. 1992. J. Periodont. Res. 27, 101.
Zimmermann, B., et al. 1992. Eur. Arch. Biol. 103, 93.
Schröter-Kermani, C., et al. 1991. Matrix 11, 428.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin V T Engebretsen et al.
Journal of applied physiology (Bethesda, Md. : 1985), 114(8), 988-997 (2013-02-16)
On the basis of the role of small, leucine-rich proteoglycans (SLRPs) in fibrogenesis and inflammation, we hypothesized that they could be involved in cardiac remodeling and reverse remodeling as occurs during aortic stenosis and after aortic valve replacement. Thus, in
Yusuke Kubo et al.
International journal of molecular sciences, 21(3) (2020-02-12)
Tenocytes are mechanosensitive cells intimately adapting their expression profile and hence, their phenotype to their respective mechanomilieu. The immunolocalization and expression intensity of tenogenic, anabolic and catabolic markers in tenocytes in response to in vitro mechanical loading have not been
Mei Li et al.
International journal of molecular medicine, 38(2), 513-520 (2016-06-18)
Hepatitis B virus (HBV)-associated glomerulo-nephritis is the most common extra-hepatic disorder occurring with hepatitis B virus infection. In the present study, we hypothesized that HBV X protein (HBx) may play a critical role in renal interstitial fibrosis, as HBx has been shown to induce epithelial-mesenchymal transition (EMT)
Kun Zhang et al.
Theranostics, 9(25), 7566-7582 (2019-11-07)
Liver fibrosis leading to cirrhosis is one of the major health burdens worldwide with currently limited therapeutic options available. Long noncoding RNAs (lncRNAs) play important roles in various biological and pathological processes in a cell- or tissue-specific manner. However, there
Eric J Suh et al.
Genome biology, 13(12), R121-R121 (2012-12-25)
Although quiescence (reversible cell cycle arrest) is a key part in the life history and fate of many mammalian cell types, the mechanisms of gene regulation in quiescent cells are poorly understood. We sought to clarify the role of microRNAs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service