Skip to Content
Merck
All Photos(1)

Key Documents

A-004-M

Sigma-Aldrich

Poly-ʟ-Ornithine

synthetic, liquid, 0.1 mg/mL, suitable for cell culture

Synonym(s):

Poly-L-Ornithine Solution (0.01%), A-004-M, Sigma-Aldrich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
eCl@ss:
32160801
NACRES:
NA.75

Product Name

Poly-L-Ornithine Solution (0.01%),

biological source

synthetic

Quality Level

needle size

2.0 mm

sterility

sterile

form

liquid

mol wt

Mw 78000 Da

manufacturer/tradename

NovaSeptum®

concentration

0.01 % (w/v)
0.1 mg/mL

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable

General description

Research Category: Stem Cell Research

Research Sub Category: Adhesion (CAMs)

Application

Poly-L-Ornithine Solution (0.01%) has been used to coat:
  • glass-bottom plates for cell plating in a research study investigating the localization of cMET in MDA-MB-468 cells using confocal microscopy
  • culture dishes for adult human neural stem cells (ahNSCs) primary culture
  • 6-well plates for induced pluripotent stem cells (iPSCs) culture and neuronal differentiation

Quality

Solution is clear with no particulates present. Solution is sterile and suitable for cell culture applications.

Physical form

Product is presented in sterile water at 0.01% w/v (0.1mg/ml). Product is filtered through a 0.2 micron filter before freezing.

Preparation Note

1. Thaw Poly-L-Ornithine solution at room temperature.

2. Dilute Poly-L-Ornithine solution to the desired concentration in sterile water.

3. Fully coat the cell culture surface with diluted Poly-L-Ornithine solution. Use 5 mL volume for 6-cm plates and 10 mL volume for 10-cm plates and T75 flasks.

4. Allow the cell culture vessel to sit at room temperature overnight.

5. Aspirate the Poly-L-Ornithine solution the following day and rinse the vessel with sterile water followed by coating with desired ECM protein. Optimal coating concentrations must be determined by the end user. Typical coating concentrations range from 10ug/ml to 100ug/ml based on cell type and application.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt. Avoid multiple freeze thaw cycles to maintain product integrity.

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of novel cMET-targeting antibody Fab drug conjugates as potential treatment for solid tumors with highly expressed cMET
Expert opinion on biological therapy, 1137-1149 (2023)
Ryan S Hippman et al.
ACS chemical neuroscience, 14(24), 4363-4382 (2023-12-10)
Autophagy is a major catabolic degradation and recycling process that maintains homeostasis in cells and is especially important in postmitotic neurons. We implemented a high-content phenotypic assay to discover small molecules that promote autophagic flux and completed target identification and
Geun-Hyoung Ha et al.
Aging, 14(22), 8944-8969 (2022-11-30)
Mismatches between pre-clinical and clinical results of stem cell therapeutics for ischemic stroke limit their clinical applicability. To overcome these discrepancies, precise planning of pre-clinical experiments that can be translated to clinical trials and the scientific elucidation of treatment mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service