8.52261
Di-tert-butyl dicarbonate
≥97.0% (GC), for peptide synthesis, Novabiochem®
Synonym(s):
Di-tert-butyl dicarbonate, Di-tert-butyl dicarbonate, Di-tert-butyl pyrocarbonate
About This Item
Recommended Products
Product Name
Di-tert-butyl dicarbonate, Novabiochem®
Quality Level
product line
Novabiochem®
Assay
≥97.0% (GC)
form
crystalline solid
autoignition temp.
460 °C
potency
>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
reaction suitability
reaction type: Coupling Reactions
manufacturer/tradename
Novabiochem®
bp
56-57 °C/0.7 hPa
mp
20-24 °C
transition temp
flash point 37 °C
density
1.02 g/cm3 at 20 °C
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
O(C(C)(C)C)C(=O)OC(=O)OC(C)(C)C
InChI
1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3
InChI key
DYHSDKLCOJIUFX-UHFFFAOYSA-N
Related Categories
General description
Analysis Note
Appearance of substance (visual): crystal to crystalline solid
Assay (GC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
98.6 °F - closed cup
Flash Point(C)
37 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service