Skip to Content
Merck
All Photos(1)

Documents

34660

Sigma-Aldrich

Di-tert-butyl dicarbonate

≥98.0% (GC), for peptide synthesis

Synonym(s):

Boc2O, Boc anhydride, Di-tert-butyl pyrocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3COCO]2O
CAS Number:
Molecular Weight:
218.25
Beilstein:
1911173
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

product name

Di-tert-butyl dicarbonate, ≥98.0% (GC)

Quality Level

Assay

≥98.0% (GC)

form

solid or liquid

refractive index

n20/D 1.409 (lit.)

bp

56-57 °C/0.5 mmHg (lit.)

mp

23 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

InChI

1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3

InChI key

DYHSDKLCOJIUFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di-tert-butyl dicarbonate (Boc2O) is a reagent mainly used for the introduction of the Boc protecting group to amine functionalities. It is also used as a dehydrating agent in some organic reactions, particularly with carboxylic acids, certain hydroxyl groups, or with primary nitroalkanes.

Automated Boc protection and deprotection can be done using Synple Automated Synthesis Platform (SYNPLE-SC002), Boc protection cartridges ((SYNPLE-B001), (SYNPLE-B002), and Boc deprotection cartridges (SYNPLE-B011)

Application

Di-tert-butyl dicarbonate (Boc2O) has been used in the synthesis of:
  • An azobenzene amino acid(aa).
  • N-tert-butoxycarbonyl-3,4-methylenedioxymethamphetamine (t-BOC-MDMA) from MDMA.

It can also be used as a reagent:
  • To introduce acid-labile Boc-protecting group in amino acids, peptides, and proteins.        
  • To prepare styrene derivatives by Heck olefination of aromatic carboxylic acids in the presence of a Pd catalyst.        
  • To synthesize oxazolidin-2-ones and imidazolidin-2-ones by isocyanation of 1,2-aminoalcohols and 1,2-diamines using DMAP as a catalyst.       
  • In the preparation of N-tert-butoxycarbonyl-3,4-methylenedioxymethamphetamine (t-Boc-MDMA) from MDMA.
  • For the conversion of amines to corresponding isocyanates, carbamates, and urea derivatives.
  • In the N-BOC-ylation of amides.
  • In the N-BOC-ylation of sensitive compounds under non-aqueous conditions.
Reagent for the introduction of the Boc protecting group.

Warning

Hydrolyzes to t-butanol and CO2; causes internal pressure in bottle if exposed to moisture.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling ephedrine prepared from N-methylalanine via the Akabori-Momotani Reaction.
Doddridge A, et al.
Drug Testing and Analysis, 10(3), 548-556 (2018)
Use of Di-tert-butyl-dicarbonate Both as a Protecting and Activating Group in the Synthesis of Dipeptides
Jhaumeer LS, et al.
Synthetic Communications, 37(23), 4191-4197 (2007)
Exhaustive tert-Butoxycarbonylation of Peptide Nitrogens
L. Grehn and Ragnarsson U
Angewandte Chemie (International Edition in English), 24(6), 510-511 (1985)
tert-BUTOXYCARBONYLATION OF AMINO ACIDS AND THEIR DERIVATIVES: N-tert-BUTOXYCARBONYL-l-PHENYLALANINE
O. Keller et al.
Organic Syntheses, 63, 160-160 (1985)
Synthesis and application of an azobenzene amino acid as a light-switchable turn element in polypeptides.
Aemissegger A & Hilvert D.
Nature Protocols, 2(1), 161-161 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service