Skip to Content
Merck
All Photos(1)

Key Documents

8.07564

Sigma-Aldrich

9,10-Phenanthrenequinone

for synthesis

Synonym(s):

9,10-Phenanthrenequinone, 9,10-Phenanthrenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O2
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-515-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (HPLC)

form

powder

autoignition temp.

630 °C (DIN 51794)

potency

>16000 mg/kg LD50, oral (Rat)

bp

>360 °C

mp

203-206 °C

transition temp

flash point 245 °C

solubility

7.5 mg/L

bulk density

350 kg/m3

storage temp.

2-30°C

SMILES string

O=[c]1c2c(c3c([c]1=O)cccc3)cccc2

InChI

1S/C14H8O2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8H

InChI key

YYVYAPXYZVYDHN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 206 °C
Melting range (upper value): ≤ 210 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

473.0 °F

Flash Point(C)

245 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service