Skip to Content
Merck
All Photos(1)

Documents

681676

Sigma-Aldrich

Wortmannin

InSolution, ≥95%, fungal metabolite that acts as a selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase)

Synonym(s):

InSolution Wortmannin, MLCK Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O8
Molecular Weight:
428.43
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

A 10 mM (1 mg/233 µl) solution of Wortmannin (Cat. No. 681675) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingya Liao et al.
Pharmaceuticals (Basel, Switzerland), 15(11) (2022-11-11)
Glucose transporter 4 (GLUT4) is a membrane protein that regulates blood glucose balance and is closely related to type 2 diabetes. Andrographolide (AND) is a diterpene lactone extracted from herbal medicine Andrographis paniculata, which has a variety of biological activities.
Yanhong Yao et al.
Pharmaceuticals (Basel, Switzerland), 16(1) (2023-01-22)
GLUT4 is an important glucose transporter, which is closely related to insulin resistance and type 2 diabetes. In this study, we investigated the mechanism of Estradiol Dipropionate (EDP) on uptake of glucose in L6 skeletal muscle cells. In our study
Mio Iwasaki et al.
iScience, 25(5), 104289-104289 (2022-05-17)
The effects of transcription factors on the maintenance and differentiation of human-induced or embryonic pluripotent stem cells (iPSCs/ESCs) have been well studied. However, the importance of posttranscriptional regulatory mechanisms, which cause the quantitative dissociation of mRNA and protein expression, has
Georgios Konstantinidis et al.
Methods in molecular biology (Clifton, N.J.), 1854, 187-195 (2018-02-23)
Autophagy is a fundamental cellular catabolic pathway mediating the recycling of cellular components. Autophagy has been implicated in pathogenesis of diverse diseases such as neurodegeneration and cancer. Due to the therapeutic potential, the autophagy-modulating agents have profoundly enriched the spectrum
Quangdon Tran et al.
Cell death & disease, 13(6), 543-543 (2022-06-11)
The selective autophagy of damaged mitochondria is called mitophagy. Mitochondrial dysfunction, mitophagy, and apoptosis have been suggested to be interrelated in various human lung carcinomas. Leucine zipper EF-hand-containing transmembrane protein-1 (LETM1) was cloned in an attempt to identify candidate genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service