Skip to Content
Merck
All Photos(1)

Key Documents

648485-M

Millipore

TrpA1 Antagonist, HC-030031

The TrpA1 Antagonist, HC-030031, also referenced under CAS 349085-38-7, controls the biological activity of TrpA1. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

TrpA1 Antagonist, HC-030031, HC-030031

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5O3
CAS Number:
Molecular Weight:
355.39
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[n]2(c3c(nc2)N(C(=O)N(C3=O)C)C)CC(=O)Nc1ccc(cc1)C(C)C

InChI

1S/C18H21N5O3/c1-11(2)12-5-7-13(8-6-12)20-14(24)9-23-10-19-16-15(23)17(25)22(4)18(26)21(16)3/h5-8,10-11H,9H2,1-4H3,(H,20,24)

InChI key

HEQDZPHDVAOBLN-UHFFFAOYSA-N

General description

A cell-permeable and selective TRPA1 antagonist that reversibly antagonizes AITC- and formalin-evoked calcium influx (IC50 = 6.2 µM and 5.3 µ M, respectively), and blocks AITC- and formalin-induced inward and outward currents in TRPA1-expressing HEK293 cells in a patch-clamp assay. It demonstrates activity towards rat and mouse TRPA1 orthologues, but does not block currents mediated by TRPV1, TRPV3, TRPV4, hERG, or NaV1.2 channels. This compound is also shown to block TRPA1 activation induced by formalin in cultured rat DRG neurons, and attenuate pain-related responses to formalin in rats (300 mg/kg, i.p.), in vivo.

Warning

Toxicity: Standard Handling (A)

Other Notes

Taylor-Clark, T., et al. 2008. Mol. Pharmacol.73, 274.
McNamara, C., et al. 2007. Proc. Natl. Acad. Sci. U S A.104, 13525.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service