Skip to Content
Merck
All Photos(1)

Key Documents

559285

Sigma-Aldrich

RSK Inhibitor, SL0101

The RSK Inhibitor, SL0101 controls the biological activity of RSK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

RSK Inhibitor, SL0101, Kaempferol-3-O-(3ʹʹ,4ʹʹ-di-O-acetyl-α-L-rhamnopyranoside), p90 Ribosomal S6 Kinase Inhibitor, SL0101

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)OC(=O)C)OC(=O)C)O)Oc2[c](c3c([o]c2c4ccc(cc4)O)cc(cc3O)O)=O

InChI

1S/C25H24O12/c1-10-21(34-11(2)26)24(35-12(3)27)20(32)25(33-10)37-23-19(31)18-16(30)8-15(29)9-17(18)36-22(23)13-4-6-14(28)7-5-13/h4-10,20-21,24-25,28-30,32H,1-3H3/t10-,20+,21-,24-,25-/m0/s1

InChI key

SXOZSDJHGMAEGZ-IGKKHSBFSA-N

General description

A cell-permeable Kaempferol (Cat. No. 420345) glycoside that targets the the N-terminal kinase domain of p90 ribosomal S6 kinase and inhibits RSK kinase activity (IC50 = 89 nM with 10 µM ATP) in a selective, reversible, and ATP-competitive (Ki = 1 µM) manner. Shown to inhibit the proliferation of MCF-7, but not the normal breast cell line MCF-10A, and specifically block PDB-induced cellular phosphorylation of RSK substrate p140, but not that of RSK or RSK upstream kinases even at concentrations as high as 100 µM.
A cell-permeable Kaempferol (Cat. No. 420345) glycoside that targets the the N-terminal kinase domain of p90 ribosomal S6 kinase and inhibits RSK kinase activity (IC50 = 89 nM with 10 µM ATP) in a selective, reversible, and ATP-competitive (Ki = 1 µM) manner. Shown to inhibit the proliferation of MCF-7, but not the normal breast cell line MCF-10A, and specifically block PDB-induced cellular phosphorylation of RSK substrate p140, but not that of RSK or RSK upstream kinases even at concentrations as high as 100 µM. Also available as a 10 mM solution in DMSO (Cat. No. 559292).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Smith, J.A., et al. 2007. Bioorg. Med. Chem.15, 5018.
Smith, J.A., et al. 2006. Bioorg. Med. Chem.14, 6043.
Xu, Y.M., et al. 2006. Bioorg. Med. Chem.14, 3974.
Maloney, D.J. and Hecht, S.M., 2005. Org. Lett.7, 1097.
Smith, J.A., et al. 2005. Cancer Res.65, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed Mahameed et al.
Biotechnology and bioengineering, 116(3), 569-580 (2018-12-05)
Protein translation has emerged as a critical bottleneck for overall productivity of biological molecules. An augmentation of protein translation can be achieved by cell line engineering or by sophisticated vector design. However, for industrial process development purposes, identification of media
Coralie Dessauges et al.
Molecular systems biology, 18(6), e10670-e10670 (2022-06-14)
Combining single-cell measurements of ERK activity dynamics with perturbations provides insights into the MAPK network topology. We built circuits consisting of an optogenetic actuator to activate MAPK signaling and an ERK biosensor to measure single-cell ERK dynamics. This allowed us

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service