Skip to Content
Merck
All Photos(1)

Key Documents

5.04817

Sigma-Aldrich

UK-5099

≥99% (HPLC), solid, mitochondrial pyruvate carrier inhibitor, Calbiochem

Synonym(s):

Mitochondrial Pyruvate Carrier Inhibitor, UK5099, ( E)-2-Cyano-3-(1-phenyl-1H-indol-3-yl)acrylic acid, ( E)-2-Cyano-3-(1-phenyl-1H-indol-3-yl)-2-propenoic acid, MPC Inhibitor, PF-1005023, (E)-2-Cyano-3-(1-phenyl-1H-indol-3-yl)acrylic acid, (E)-2-Cyano-3-(1-phenyl-1H-indol-3-yl)-2-propenoic acid, MPC Inhibitor, PF-1005023

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$54.72
5 G
$219.00

$54.72

List Price$62.90Save 13%

In StockDetails


To order products, please contact your local dealer.

Select a Size

Change View
1 G
$54.72
5 G
$219.00

About This Item

Empirical Formula (Hill Notation):
C18H12N2O2
CAS Number:
Molecular Weight:
288.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$54.72

List Price$62.90Save 13%

In StockDetails


To order products, please contact your local dealer.

Product Name

Mitochondrial Pyruvate Carrier Inhibitor, UK5099,

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

bright yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[n]2(c3c(c(c2)\C=C(\C(=O)O)/C#N)cccc3)c1ccccc1

General description

A cell-permeable α-cyanocinnamate compound that acts as a rapid, active site thiol modifying, time-dependent and non-competitive inhibitor of bi-directional pyruvate transport across plasma membranes in yeast, bacteria, Drosophila, and humans (IC50 = 50 nM in rat heart mitochondria; Ki ≤ 1 µM) with excellent selectivity over monocarboxylates and anions. Shown to significantly decrease pyruvate-driven respiration, enhance glucose uptake and activate AMPK (2 µM in human myocytes and skeletal muscle myotubes).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
mitochondria
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C or -70°C). Stock solutions are stable for up to 2 weeks at -20°C or for up to 6 weeks at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Divakaruni, A.S., et al. 2013. Proc. Natl. acad. Sci. USA110, 5422.
Bricker, D.K., et al. 2012. Science337, 96.
Herzig, S., et al. 2012. Science337, 93.
Hinoi, E., et al. 2006. Mol. Pharmacol.70, 925.
Hildyard, J.C.W., et al. 2005. Biochim. Biophys. Acta1707, 221.
Halestrap, A.P. 1978. Biochem. J.172, 377.
Halestrap, A.P. 1975. Biochem. J.145, 85.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service