Skip to Content
Merck
All Photos(1)

Key Documents

487928

Sigma-Aldrich

Nocodazole

≥99% (HPLC), solid, microtuble inhibitor, Calbiochem

Synonym(s):

Nocodazole, R17934, Methyl-[5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3O3S
CAS Number:
Molecular Weight:
301.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Nocodazole, Inhibitor of mitosis.

Quality Level

description

RTECS - DD6521000

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[s]1c(ccc1)C(=O)c2cc3nc([nH]c3cc2)NC(=O)OC

InChI

1S/C14H11N3O3S/c1-20-14(19)17-13-15-9-5-4-8(7-10(9)16-13)12(18)11-3-2-6-21-11/h2-7H,1H3,(H2,15,16,17,19)

InChI key

KYRVNWMVYQXFEU-UHFFFAOYSA-N

General description

Inhibitor of mitosis. Has highly specific antimicrotubular activity for mammalian cells in culture. Promotes tubulin depolymerization. Also available as a 50 mM solution in DMSO (Cat. No. 487929).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Promotes tubulin depolymerization
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 1 year at -20°C.

Other Notes

Park, H., et al. 2011. Chem. Med. Chem.in press.
Bunz, F., et al. 1998. Science 282, 1497.
Fleischman, L.F., et al. 1993. Oncogene8, 771.
Pomies, P., and Block, M.R. 1992. Biochem. Biophys. Res. Commun.189, 1429.
Rennison, M.E., et al. 1992. J. Cell Sci. 102, 239.
Thyberg, J., and Moskalewski, S. 1992. J. Cell Sci.103, 1167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hieng Chiong Tie et al.
Methods in molecular biology (Clifton, N.J.), 2557, 113-125 (2022-12-14)
The Golgi complex is essential for protein transport and posttranslational modification in mammalian cells. It is critical to know the cisternal distribution of Golgi proteins to understand Golgi functions. The cis-to-trans or axial localization of a Golgi protein can be
Nicolás Cifuentes-Muñoz et al.
Journal of virology, 91(24) (2017-10-06)
Human metapneumovirus (HMPV) causes significant upper and lower respiratory disease in all age groups worldwide. The virus possesses a negative-sense single-stranded RNA genome of approximately 13.3 kb encapsidated by multiple copies of the nucleoprotein (N), giving rise to helical nucleocapsids.
Archan Ganguly et al.
Neuron, 109(18), 2884-2901 (2021-09-18)
In non-neuronal cells, clathrin has established roles in endocytosis, with clathrin cages enclosing plasma membrane infoldings, followed by rapid disassembly and reuse of monomers. However, in neurons, clathrin is conveyed in slow axonal transport over days to weeks, and the
Esra Unsal et al.
Chemical biology & drug design, 88(6), 783-794 (2016-11-05)
Aurora B is a serine/threonine kinase that has a central role in the regulation of mitosis. The observation of Aurora B overexpression in cancer makes it a promising target to develop antitumoral inhibitors. We describe a new potential inhibitor that
Benoit Dehapiot et al.
PLoS biology, 19(9), e3001376-e3001376 (2021-09-08)
Mammalian oocyte meiotic divisions are highly asymmetric and produce a large haploid gamete and 2 small polar bodies. This relies on the ability of the cell to break symmetry and position its spindle close to the cortex before anaphase occurs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service