Skip to Content
Merck
All Photos(1)

Key Documents

445800

Sigma-Aldrich

Merbarone

≥95% (HPLC), solid, topoisomerase II inhibitor, Calbiochem®

Synonym(s):

Merbarone, 5-(N-Phenylcarboxamido)-2-thiobarbituric Acid, NSC-336628

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N3O3S
CAS Number:
Molecular Weight:
263.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Merbarone, A cell-permeable anticancer drug that inhibits the catalytic activity of topoisomerase II (topo II) without damaging DNA or stabilizing DNA-topo II cleavable complexes.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

faint pink

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Sc1nc(c(c(n1)O)C(=O)Nc2ccccc2)O

InChI

1S/C11H9N3O3S/c15-8(12-6-4-2-1-3-5-6)7-9(16)13-11(18)14-10(7)17/h1-5H,(H,12,15)(H3,13,14,16,17,18)

InChI key

GFYRZTLCYQQVHZ-UHFFFAOYSA-N

General description

A cell-permeable anticancer drug that inhibits the catalytic activity of topoisomerase II (topo II) without damaging DNA or stabilizing DNA-topo II cleavable complexes (IC50 = 20 µM for purified mammalian topo II versus IC50 = ~ 200 µM for topo I). Also induces apoptosis in human leukemic CEM cells through a caspase-3-like protease-dependent mechanism.
A cell-permeable, anticancer drug that inhibits the catalytic activity of DNA topoisomerase II (topo II) without damaging DNA or stabilizing DNA-topo II cleavable complexes (IC50 = 20 µM for purified mammalian topoisomerase II versus IC50 ~200 µM for topoisomerase I). Also induces apoptosis in human leukemic CEM cells through a caspase-3-like protease-dependent mechanism.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Catalytic activity of topoisomerase II
Product does not compete with ATP.
Reversible: no
Target IC50: 20 µM for purified topoisomerase II (topo II)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Khelifa, T., and Beck, W.T. 1999. Mol. Pharmacol. 55, 548.
Drake, F.H., et al. 1989. Cancer Res.49, 2578.
Brewer, A.D., et al. 1985. Biochem. Pharmacol.34, 2047.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nootan Pandey et al.
The Journal of cell biology, 219(1) (2019-11-13)
Topoisomerase II (Topo II) is essential for mitosis since it resolves sister chromatid catenations. Topo II dysfunction promotes aneuploidy and drives cancer. To protect from aneuploidy, cells possess mechanisms to delay anaphase onset when Topo II is perturbed, providing additional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service