B0936
BRL 50481
≥98% (HPLC), solid
Synonym(s):
5-Nitro-2,N,N-trimethylbenzenesulfonamide
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
color
white to off-white
solubility
DMSO: >10 mg/mL
H2O: insoluble
originator
GlaxoSmithKline
storage temp.
2-8°C
SMILES string
CN(C)S(=O)(=O)c1cc(ccc1C)[N+]([O-])=O
InChI
1S/C9H12N2O4S/c1-7-4-5-8(11(12)13)6-9(7)16(14,15)10(2)3/h4-6H,1-3H3
InChI key
IFIUFCJFLGCQPH-UHFFFAOYSA-N
Gene Information
human ... PDE7A(5150) , PDE7B(27115)
Biochem/physiol Actions
Features and Benefits
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service