Skip to Content
Merck
All Photos(1)

Documents

220551

Sigma-Aldrich

Chloramphenicol

Chloramphenicol, CAS 56-75-7, is a synthetic bacteriostatic antibiotic that inhibits the translation of RNA by blocking the peptidyltransferase reaction on ribosomes.

Synonym(s):

Chloramphenicol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12Cl2N2O5
CAS Number:
Molecular Weight:
323.13
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (by Assay)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

gray to off-white

solubility

water: 2.5 mg/mL
methanol: 35 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)

InChI key

WIIZWVCIJKGZOK-UHFFFAOYSA-N

General description

Synthetic bacteriostatic antibiotic that inhibits the translation of RNA by blocking the peptidyltransferase reaction on ribosomes.
Synthetic bacteriostatic antibiotic that inhibits the translation of RNA to protein by binding to the 50S ribosomal subunit, blocking the peptidyl transferase reaction. Chloramphenicol resistance is encoded by the resistance gene cat, whose product, chloramphenicol acetyltransferase, inactivates the antibiotic via acetylation. Typically used at 30-50 µg/ml for selection of chloramphenicol-resistant Escherichia coli.

Biochem/physiol Actions

Potency: ≥970 µg/mg
Primary Target
trasnslation of RNA

Other Notes

Lech, K., and Brent, R. 1995. in Current Protocols in Mol. Biol. Vol. 1, (Ausubel, F.M., et al., Eds.) John Wiley & Sons, New York.
Lu, J., and Jiang, C. 1993. Biochem. Biophys. Res. Commun.196, 12.
Saltarelli, M.J., et al. 1993. Virol.197, 35.
Holt, J.T. 1992. Ann. N.Y. Acad. Sci.660, 88.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition. Cold Spring Harbor, NY, p. 1.6, A.6.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrés Aranda-Díaz et al.
eLife, 9 (2020-01-30)
Predicting antibiotic efficacy within microbial communities remains highly challenging. Interspecies interactions can impact antibiotic activity through many mechanisms, including alterations to bacterial physiology. Here, we studied synthetic communities constructed from the core members of the fruit fly gut microbiota. Co-culturing
Leslie Ann Jaramillo Koyama et al.
PLoS biology, 18(1), e3000567-e3000567 (2020-01-28)
Cell- and tissue-level processes often occur across days or weeks, but few imaging methods can capture such long timescales. Here, we describe Bellymount, a simple, noninvasive method for longitudinal imaging of the Drosophila abdomen at subcellular resolution. Bellymounted animals remain
Heidi A Arjes et al.
Current biology : CB, 30(6), 1011-1022 (2020-02-16)
The presence or absence of oxygen in the environment is a strong effector of cellular metabolism and physiology. Like many eukaryotes and some bacteria, Bacillus subtilis primarily utilizes oxygen during respiration to generate ATP. Despite the importance of oxygen for
Maxime Louet et al.
eLife, 10 (2021-09-04)
There is increasing support for water molecules playing a role in signal propagation through G protein-coupled receptors (GPCRs). However, exploration of the hydration features of GPCRs is still in its infancy. Here, we combined site-specific labeling with unnatural amino acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service