Skip to Content
Merck
All Photos(1)

Key Documents

A0166

Sigma-Aldrich

Ampicillin sodium salt

powder or crystals, BioReagent, suitable for cell culture

Synonym(s):

D-(−)-α-Aminobenzylpenicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3NaO4S
CAS Number:
Molecular Weight:
371.39
Beilstein:
4119211
EC Number:
MDL number:
UNSPSC Code:
51281716
PubChem Substance ID:
NACRES:
NA.76

biological source

synthetic

product line

BioReagent

form

powder or crystals

potency

845-988 μg per mg (C16H18N3O4S, Calculated on the anhydrous basis)

technique(s)

cell culture | mammalian: suitable

color

white to off-white

mp

215 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].CC1(C)SC2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ampicillin sodium salt, belonging to the extended-spectrum β-lactam family, stands as a semisynthetic derivative of penicillin with versatile applications as a broad-spectrum antibiotic. It exerts inhibitory effects on bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), thereby impeding peptidoglycan synthesis—an integral process in the formation of the bacterial cell wall. This antibiotic demonstrates activity against an extensive spectrum of bacteria, encompassing both Gram-positive and Gram-negative strains such as E. coli, β-lactam sensitive vancomycin-resistant Enterococcus (VRE), Staphylococcus aureus, and Streptococcus pneumoniae.

In research, ampicillin plays a pivotal role in microbiological, biochemical, and cell culture investigations. Its utilization in laboratories extends to studying antibiotic resistance and penetration limitations, exploring the synergistic interactions between multiple antibiotics, and serving as a crucial component for the selection and maintenance of recombinant plasmids in E. coli. Through these applications, ampicillin sodium salt contributes significantly to advancing the understanding of antibiotic efficacy, bacterial responses, and molecular processes, making it an indispensable tool in various facets of scientific research.

Application

Ampicillin is a semi-synthetic derivative of penicillin that functions as a broad-spectrum antibiotic. It has been used to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, certain bloodstream infections, and has been used to develop PCR assays to detect resistance genes in cerebrospinal fluid.

Recommended for antibacterial use in cell culture media at 100 mg/L.
Recommended for use in ampicillin-resistance studies at 20-125 μg/ml.

Biochem/physiol Actions

Mode of Action: This is a ß-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

Mode of Resistance: Administration with ß-lactamase cleaves the ß-lactam ring of Ampicillin and inactivates it.

Antimicrobial Spectrum: Includes both gram-positive (similar to benzylpenicillin) and gram-negative bacteria (similar to tetracyclines and chloramphenicol.

Features and Benefits

  • High quality antibiotic suitable for multiple research applications
  • Broad-spectrum antibiotic
  • Inhibits bacterial cell-wall synthesis
  • Active against Gram-positive and Gram-negative bacteria
  • Commonly used in Cell Culture, Cell Biology and Biochemical research

Packaging

5g, 25g, 100g

Caution

This product has been reported stable as supplied at 25°C at 43% and 81% relative humidity for six weeks. It is stable at 37°C for three days. Additional studies have shown that the stability of Ampicillin in solution is a function of pH, temperature and the identity of the buffer. It′s activity is quickly lost when stored above pH 7. Optimal storage conditions are suggested as 2-8°C, and pH 3.8-5 where its activity was retained at 90%+ for a week.

Preparation Note

Ampicillin is reported as slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils but soluble in dilute acids or bases. The solution should not be autoclaved; a stock solution should be sterilized through filtration and stored frozen, where it will be stable for months.

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

comparable product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ashley Sobran et al.
Journal of bacteriology, 201(17) (2019-06-27)
To detect and respond to the diverse environments they encounter, bacteria often use two-component regulatory systems (TCS) to coordinate essential cellular processes required for survival. In pathogenic Bordetella species, the BvgAS TCS regulates expression of hundreds of genes, including those
Daejin Lim et al.
Frontiers in bioengineering and biotechnology, 8, 840-840 (2020-08-09)
Targeted delivery of drugs is a key aspect of the successful treatment of serious conditions such as tumors. In the pursuit of accurate delivery with high specificity and low size limit for peptide drugs, a synthetic type 3 secretion system
Alexander Frank et al.
Current biology : CB, 28(16), 2597-2606 (2018-08-07)
Synchronization of circadian clocks to the day-night cycle ensures the correct timing of biological events. This entrainment process is essential to ensure that the phase of the circadian oscillator is synchronized with daily events within the environment [1], to permit
Nuria Fernández-Hidalgo et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1261-1268 (2013-02-09)
The aim of this study was to compare the effectiveness of the ampicillin plus ceftriaxone (AC) and ampicillin plus gentamicin (AG) combinations for treating Enterococcus faecalis infective endocarditis (EFIE). An observational, nonrandomized, comparative multicenter cohort study was conducted at 17
Wei Lin et al.
Molecular cell, 70(1), 60-71 (2018-04-03)
Fidaxomicin is an antibacterial drug in clinical use for treatment of Clostridium difficile diarrhea. The active ingredient of fidaxomicin, lipiarmycin A3 (Lpm), functions by inhibiting bacterial RNA polymerase (RNAP). Here we report a cryo-EM structure of Mycobacterium tuberculosis RNAP holoenzyme in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service