Skip to Content
Merck
All Photos(1)

Key Documents

1.93017

Supelco

L(+)-Tartaric acid

for analysis EMPARTA® ACS

Synonym(s):

L-(+)-Tartaric acid, (2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
MDL number:
UNSPSC Code:
12352106

grade

ACS reagent

vapor density

5.18 (vs air)

autoignition temp.

797 °F

mp

170-172 °C (lit.)

storage temp.

room temp

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93017.0521: 500 g, Minimum order quantity: 20
1.93017.5021: 5 kg, Minimum order quantity: 4

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service