Skip to Content
Merck
All Photos(1)

Key Documents

W304409

Sigma-Aldrich

L-(+)-Tartaric acid

≥99.7%, FCC, FG

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
FEMA Number:
3044
Beilstein:
1725147
EC Number:
Council of Europe no.:
18
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.018
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 163.110
FDA 21 CFR 163.111
FDA 21 CFR 163.112
FDA 21 CFR 184.1099

vapor density

5.18 (vs air)

Assay

≥99.7%

form

crystalline powder

optical activity

[α]20/D +12.5°, c = 20 in H2O

autoignition temp.

797 °F

mp

170-172 °C (lit.)

solubility

water: soluble 150 g/L at 25 °C

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
heavy metals (as Pb): ≤2 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

L-(+)-Tartaric acid is an organic acid commonly used as an acidulant, flavor enhancer, firming agent and humectant.

Application


  • Terahertz-spectroscopy for non-destructive determination of crystallinity of L-tartaric acid in smartFilms and tablets made from paper.: This study leverages terahertz spectroscopy to assess the crystallinity of L-(+)-tartaric acid in innovative pharmaceutical applications, enhancing non-destructive testing methods for quality control (Ornik et al., 2020 May). Link to the article.

  • Enhanced pulmonary absorption of poorly soluble itraconazole by micronized cocrystal dry powder formulations.: Research shows the use of L-(+)-tartaric acid in cocrystal formulations with itraconazole to improve its pulmonary absorption, demonstrating a significant advancement in drug delivery technologies (Karashima et al., 2017 Jun). Link to the article.

  • Physicochemical Evaluation and Developability Assessment of Co-amorphouses of Low Soluble Drugs and Comparison to the Co-crystals.: This article discusses the role of L-(+)-tartaric acid in enhancing the solubility and bioavailability of pharmaceuticals through co-amorphous systems, offering a critical insight into drug formulation strategies (Yamamoto et al., 2016 Dec). Link to the article.

  • Functionalized polycarbonate derived from tartaric acid: enzymatic ring-opening polymerization of a seven-membered cyclic carbonate.: This research explores the synthesis of biodegradable polymers from L-(+)-tartaric acid, emphasizing its utility in developing environmentally friendly materials (Wu et al., 2008 Oct). Link to the article.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 1794-1794 (2012)
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The
Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single
Jianping Yang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(43), 13642-13650 (2012-09-22)
In this paper, we report a facile one-step hydrothermal method to synthesize phase-, size-, and shape-controlled carboxyl-functionalized rare-earth fluorescence upconversion phosphors by using a small-molecule binary acid, such as malonic acid, oxalic acid, succinic acid, or tartaric acid as capping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service