Skip to Content
Merck
All Photos(4)

Documents

17-10264

Sigma-Aldrich

ChIPAb+ JHDM1B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

JMJD, Lysine (K)-specific demethylase 2B, F-box protein FBL10, [Histone-H3]-lysine-36 demethylase 1B, JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein, F-box and leucine-rich repeat protein 10, Protein containing CXXC domain 2, Ju

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

rabbit

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

horse, mouse, pig, human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), equine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rat (based on 100% sequence homology), rabbit (based on 100% sequence homology)

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... KDM2B(84678)

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+JHDM1B set includes the JHDM1B antibody, a Normal Rabbit IgG, and control primers which amplify a 110 bp region of ChIP Primers, human β-globin. The JHDM1B and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of JHDM1B -associated chromatin.
JHDM1B (jumonji domain-containing histone demethylation protein 1B) and KDM2B. Due to the presence of a F-box motif, this protein is also known as the F-box/LRR-repeat protein 10 (FBXL 10). This protein functions as a histone demethylase for lysine 36 of histione H3 and preferentially demethylates dimethylated lysine 3 with weak or no activity for mono- and tri-methylted H3 lysine 36.

Immunogen

KLH-conjugated linear peptide corresponding to human JHDM1B.

Application

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µL of either Normal Rabbit IgG or Anti-JHDM1B and the Magna ChIP A Kit (Cat. # 17-610). Successful immunoprecipitation of JHDM1B associated DNA fragments was verified by qPCR using ChIP Primers, human β-globin as a positive locus, and β-actin promoter primers as a negative locus. (Figure 2). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
HeLa nuclear extract was probed with Anti-JHDM1B (1 ug/mL). Proteins were visualized using a Donkey Anti-Rabbit IgG secondary antibody conjugated to HRP and a chemiluminescence detection system.
Arrow indicates JHDM1B (~165 kDa). (Figure 3). An uncharacterized band appears at ~109 kDa in some lysates.
Immunocytochemistry Analysis:
Representative lot data.
Confocal fluorescent analysis of NIH/3T3 and HeLa cells using Anti-JHDM1B (Red). Actin filaments have been labeled with Alexa Fluor 488 dye -Phalloidin (Green). Nucleus is stained with DAPI (Blue). This antibody positively stains the nucleus. (Figure 4).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This ChIPAb+ JHDM1B -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: 1.4 μg (2 μL) of antibody per chromatin immuno-precipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µL of either Normal Rabbit IgG or Anti-JHDM1B and the Magna ChIP® A Kit (Cat. # 17-610). Successful immunoprecipitation of JHDM1B associated DNA fragments was verified by qPCR using ChIP Primers, human β-globin (Figure 1).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~165 kDa observed. An uncharacterized lower molecular weight band may be observed in some lysates.

Physical form

Affinity purified
Components:
Anti-JHDM1B (Rabbit Polyclonal). One vial containing 50 µL of purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl and 0.05% sodium azide before the addition of glycerol to 30%. Store at -20°C.
Concentration: 0.7 mg/mL
Normal Rabbit IgG. One vial containing 125 µg of Rabbit IgG in 125 µL of storage buffer containing 0.05% sodium azide. Store at -20°C.
ChIP Primers, human β-globin. One vial containing 75 μL of each primer (5 μM) specific for the human β-globin promoter. Store at -20°C.
FOR: AGG ACA GGT ACG GCT GTC ATC
REV: TTT ATG CCC AGC CCT GGC TC

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes normal rabbit IgG and primers specific for human β-globin.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lidia Chellini et al.
Oncogene, 43(4), 225-234 (2023-11-29)
Ewing sarcomas (ES) are aggressive paediatric tumours of bone and soft tissues. Resistance to chemotherapy and high propensity to metastasize remain the main causes of treatment failure. Thus, identifying novel targets for alternative therapeutic approaches is urgently needed. DNA/RNA helicases
Shuanghong Zhu et al.
Science advances, 10(16), eadi1782-eadi1782 (2024-04-17)
Mutant isocitrate dehydrogenases (IDHs) produce R-2-hydroxyglutarate (R-2HG), which inhibits the growth of most acute myeloid leukemia (AML) cells. Here, we showed that necroptosis, a form of programmed cell death, contributed to the antileukemia activity of R-2HG. Mechanistically, R-2HG competitively inhibited
Polycomb group and SCF ubiquitin ligases are found in a novel BCOR complex that is recruited to BCL6 targets.
Gearhart, MD; Corcoran, CM; Wamstad, JA; Bardwell, VJ
Molecular and cellular biology null
IPO8 and FBXL10: new reference genes for gene expression studies in human adipose tissue.
Hurtado del Pozo, C; Calvo, RM; Vesperinas-Garcia, G; Gomez-Ambrosi, J; Fruhbeck et al.
Obesity (Silver Spring, Md.) null
Eva J Schaefer et al.
Blood cancer discovery, 3(2), 116-135 (2022-01-12)
Polycomb repressive epigenetic complexes are recurrently dysregulated in cancer. Unlike polycomb repressive complex 2 (PRC2), the role of PRC1 in oncogenesis and therapy resistance is not well-defined. Here, we demonstrate that highly recurrent mutations of the PRC1 subunits BCOR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service