Skip to Content
Merck
All Photos(1)

Key Documents

1.18789

Supelco

Hydrogen Peroxide Test, photometric

photometric, 0.015-6.00 mg/L (H2O2), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Hydrogen Peroxide Test, photometric, 0.015-6.00 mg/L (H2O2), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

hydrogen peroxide

measuring range

0.015-6.00 mg/L (H2O2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Neocuproin)

storage temp.

15-25°C

General description

Method: photometric 0.015 - 6.00 mg/l H₂O₂ Spectroquant®This Spectroquant® Hydrogen Peroxide Reagent Test allows the accurate quantification of the hydrogen peroxide concentration in disinfectant and rinsing solutions. Method applied: In the presence of a phenanthroline derivative hydrogen peroxide reduces copper(II) ions to copper(I) ions. In the process these form an orange-colored complex which is determined photometrically. The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet. All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service