Skip to Content
Merck
All Photos(1)

Key Documents

1.16731

Supelco

Peroxide Test Strips, reflectometric

100-1000 mg/L (H₂O₂), REFLECTOQUANT®

Synonym(s):

Peroxide chemical test strips, Peroxide test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Peroxide Test, reflectometric, 100-1000 mg/L (H2O2), Reflectoquant®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: enzymatic reaction
reagent type: oxidant

measuring range

100-1000 mg/L (H2O2)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 100 - 1000 mg/l H₂O₂ Reflectoquant®
Peroxide is an all purpose disinfectant which, due to its excellent antiviral, bactericidal and fungicidal action, is widely used in the food industry. Here, the Reflectoquant test strips are suited for the determination of inorganic peroxides in aqueous solutions. Furthermore the test can be used in disinfecting and rinsing solutions (e.g. in food technology, laundries) and in swimming-pool water. Peroxidase transfers peroxide oxygen to an organic redox indicator. This produces a yellow-brown oxidation product that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the peroxide concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for peroxide.

Application


  • Food Quality Control: In the food industry, peroxide tests are used to determine the peroxide value in vegetable oils and other edible fats. This application is crucial for assessing the freshness and shelf life of food products. A recent study demonstrated the use of paper-based analytical devices for the colorimetric determination of peroxide values in vegetable oils, highlighting the simplicity and effectiveness of this method for routine food quality testing (Ghohestani et al., Food Chemistry, 2023).

Analysis Note

function control: conforms
Assay
- standard 1:
175 - 225 mg/l
- standard 2: 450 - 550 mg/l
- standard 3: 720 - 880 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service