Skip to Content
Merck
All Photos(4)

Key Documents

1.09718

Supelco

Petroleum

for analysis EMSURE®

Synonym(s):

Petroleum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352401
NACRES:
NB.21
grade:
reagent grade
vapor pressure:
<1 hPa ( 20 °C)

grade

reagent grade

Quality Level

vapor pressure

<1 hPa ( 20 °C)

product line

EMSURE®

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6-7 % (v/v)

dilution

(for analytical testing)

impurities

≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000002% Cu (Copper)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.0002 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.01% Water
≤0.2% Aromatics (as naphthalene)

kinematic viscosity

1.23 cSt(40 °C)

mp

<-50 °C

transition temp

flash point 62 °C

solubility

insoluble <0.001 g/L

density

0.749-0.761 g/cm3 at 25 °C

cation traces

Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Asp. Tox. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service