Skip to Content
Merck
All Photos(1)

Key Documents

86460

Sigma-Aldrich

White spirit

~17% aromatics basis

Synonym(s):

Mineral Spirit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
bp:
150-200 °C

form

liquid

concentration

~17% (aromatics)

refractive index

n20/D 1.437

bp

150-200 °C

density

0.78 g/mL at 20 °C

Application

White spirit is generally used:
  • As a solvent for the extraction of oil-bituminous rock organic components from mineral.
  • As a component in the mixture of organic liquids to clean the coal samples by float/sink gravity separations.


White spirit with ~17% aromatics basis can be used as a solvent in the synthesis of alkyd resin from camelina oil.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

>86.0 °F - (External MSDS)

Flash Point(C)

> 30 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of organic liquids on coking properties of a higher-inert Western Canadian coal.
Holuszko ME, et al.
Fuel Processing Technology, 155(1), 225-231 (2017)
Solvent exposure in construction and maintenance painting.
Riala R, et al.
Forensic Science International, 267, 263-266 (1984)
Synthesis and characterization of alkyd resins based on Camelina sativa oil and polyglycerol.
Nosal H, et al.
Progress in Organic Coatings, 86(1), 59-70 (2015)
Investigation of a Mechanism for Extraction of Organic Components of a Mineral Part from Imankara Field Oil-Bituminous Rocks by Ultrasonic Treatment Method in the Presence of Surfactants and Flocculants.
Beysenbayev OK and Esirkepova M
Orient. J. Chem., 34(1), 482-492 (2018)
Towards chemical profiling of ignitable liquids with comprehensive two-dimensional gas chromatography: Exploring forensic application to neat white spirits.
Sampat AAS, et al.
Forensic Science International, 267, 183-195 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service