1.08131
Sulfuric acid
96%, for the determination of viscosity acc. to DIN EN ISO 307
Synonym(s):
Sulfuric acid
About This Item
Recommended Products
Product Name
Sulfuric acid 96%, for the determination of viscosity acc. to DIN EN ISO 307
vapor density
<0.3 (25 °C, vs air)
Quality Level
vapor pressure
0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)
Assay
95.85-96.15% (alkalimetric)
form
liquid
pH
0.3 (25 °C, 49 g/L in H2O)
bp
~290 °C (lit.)
mp
-20 °C
density
1.840 g/mL at 25 °C (lit.)
storage temp.
no temp limit
SMILES string
OS(O)(=O)=O
InChI
1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)
InChI key
QAOWNCQODCNURD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service