Skip to Content
Merck
All Photos(1)

Documents

1.00403

Supelco

Molybdenum(VI) oxide

for analysis EMSURE®

Synonym(s):

Molybdenum(VI) oxide, Molybdenum trioxide, Molybdic acid anhydrous, Molybdic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MoO3
CAS Number:
Molecular Weight:
143.94
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-204-7
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.5% (precipitation titration)

form

solid

potency

2689 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

795 °C

solubility

0.49 g/L

density

4.70 g/cm3 at 26 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.002%
phosphate, silicate, arsenate (as PO43-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.0005%
NH4+: ≤0.005%
Pb: ≤0.001%

storage temp.

2-30°C

InChI

1S/Mo.3O

InChI key

JKQOBWVOAYFWKG-UHFFFAOYSA-N

Analysis Note

Assay (precipitative titration): ≥ 99.5 %
Chloride (Cl): ≤ 0.002 %
Phosphate, Arsenate, Silicate (as PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.005 %
Pb (Lead): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service