Skip to Content
Merck
All Photos(2)

Documents

1.05590

Supelco

Sodium hydroxide solution about 32%

(for the determination of nitrogen) for analysis EMSURE®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

vapor pressure

3 mmHg ( 37 °C)
8 hPa ( 20 °C)

Quality Level

product line

EMSURE®

pH

14 (20 °C in H2O)

mp

9 °C

density

1.35 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Long-term durability of discarded cork-based composites obtained by geopolymerization.: This study utilizes a 32% sodium hydroxide solution to enhance the geopolymerization of discarded cork-based composites, demonstrating its effectiveness in improving long-term durability and environmental sustainability of building materials (Poggetto GD et al., 2024).

  • Lactose-6-phosphate as an alternative to disodium phosphate in the production of processed cheese food.: Research highlights the application of a 32% sodium hydroxide solution in dairy processing, specifically in adjusting pH to optimize the use of lactose-6-phosphate as a stabilizer in processed cheese, showcasing its potential in food science (Alsaleem KA et al., 2024).

  • Production of chitosan from shrimp shell using ultrasound followed by subcritical water hydrolysis.: This publication discusses the use of a 32% sodium hydroxide solution in the extraction and purification of chitosan from shrimp shells, using advanced techniques like ultrasound and subcritical water hydrolysis, underscoring its crucial role in biopolymer production (Matouri M et al., 2024).

  • [Determination of boric acid and silicic acid in mineral water by nonsuppressed ion chromatography].: The use of a 32% sodium hydroxide solution is explored for the ion chromatographic analysis of boric and silicic acids in mineral water, reflecting its importance in analytical chemistry and environmental monitoring (Yang Z et al., 2023).

  • Lanthanum and magnesium activated palygorskite for adsorption of phosphate in piggery wastewater.: This research employs a 32% sodium hydroxide solution to activate palygorskite for phosphate adsorption in wastewater treatment, demonstrating its effectiveness in improving water treatment technologies (Zhang X et al., 2023).

Analysis Note

Assay (acidimetric,NaOH): ≥ 32.0 %
Carbonate (as Na₂CO₃): ≤ 0.2 %
Chloride (Cl): ≤ 0.004 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.003 %
Total nitrogen (N): ≤ 0.0001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service