Skip to Content
Merck
All Photos(3)

Key Documents

1.04871

SAFC

Potassium dihydrogen phosphate

crystalline, EMPROVE® ESSENTIAL, Ph. Eur., BP, JPC, NF, E 340

Pharma Manufacturing

Synonym(s):

Potassium phosphate monobasic, mono-Potassium orthophosphate, Potassium biphosphate, Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
12352302
E Number:
E340
EC Index Number:
231-913-4

Agency

BP
JPC
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

Assay

≥99.0% dry basis (alkalimetric)
98.0-100.5% (alkalimetric)

form

crystalline
solid

pH

4.2-4.6 (20 °C, 20 g/L in H2O)

mp

252.6 °C (lit.)

solubility

208 g/L

density

2.338 g/mL at 25 °C (lit.)

application(s)

liquid formulation
ophthalmics
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium hydrogen phosphate is a commonly used ingredient in both food and pharmaceutical applications. In pharmaceutical applications, potassium hydrogen phosphate is primarily used as a buffering agent.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service