Skip to Content
Merck

103773

SAFC

Benzonase® endonuclease Safety Plus

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Endonuclease from Serratia marcescens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

Serratia marcescens

Quality Level

recombinant

expressed in E. coli

sterility

non-sterile

product line

EMPROVE® EXPERT

Assay

≥99% (SDS-PAGE)

form

buffered aqueous glycerol solution

feature

autoclavable: no

concentration

≥250 units/μL

impurities

≤0.25 EU/kU bacterial endotoxins

application(s)

pharma/biopharma processes

shipped in

dry ice

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

General description

Benzonase® Nuclease is a genetically engineered endonuclease from Serratia marcescens. It degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity. It is effective over a wide range of conditions and possesses an exceptionally high specific activity. The enzyme completely digests nucleic acids to 5′-monophosphate terminated oligonucleotides (below the hybridization limit), which is ideal for removal of nucleic acids from recombinant proteins, enabling compliance with FDA guidelines for nucleic acid contamination. The ability of Benzonase to rapidly hydrolyze nucleic acids makes the enzyme an excellent choice for viscosity reduction to reduce processing time and increase yields of protein.

The enzyme consists of two subunits of 30 kDa each. It is functional between pH 6 and 10 and from 0-42°C and requires 1-2 mM Mg2+ for activation. The enzyme is also active in the presence of ionic and non-ionic detergents, reducing agents, PMSF (1 mM), EDTA (1 mM) and urea (relative activity depends on specific conditions). Activity is inhibited by <150 mM monovalent cations, <100 mM phosphate, <100 mM ammonium sulfate, or <100 mM guanidine HCl.
Benzonase® endonuclease Safety Plus Emprove® Expert is a companion product to our existing Benzonase® portfolio. All Benzonase® endonuclease products share the same amino acid sequence (proven by LC-MS/MS mass spectrometry), robustness and activity. Acting as endonuclease, degrading both DNA and RNA to small 3–5 base pairs (<6 kDa) fragments with no base preference, they are the ideal tool for nucleic acid removal in virus vector and vaccine manufacturing as demanded by the regulators. The use of Benzonase® endonuclease additionally increases the yield in virus purification, protects the downstream chromatography and filter devices from fouling and reduces feed stream viscosity. With its new features, Benzonase® endonuclease Safety Plus Emprove® Expert is ideally suited for manufacturing of cell and gene therapy agents such as adeno associated and lenti viruses, oncolytic viruses and viral vector vaccines.
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

DNA clearance for purification of biopharmaceuticals, viral vaccines, viral vector vaccines, viral vectros for cell and gene therapy applications as well as oncolytic viruses.
Used for the removal of nucleic acid from protein samples.

Biochem/physiol Actions

Digests native or heat-denatured DNA and RNA.

Packaging

  • 100,000 Units
  • 500,000 Units
  • 5,000,000 Units + 50,000 Units tailgate sample

Preparation Note

Benzonase® endonuclease Safety Plus Emprove® Expert is manufactured under good manufacturing practices (GMP ICH Q7)

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service