Skip to Content
Merck
All Photos(1)

Documents

U4883

Supelco

Urea

8 M (after reconstitution with 16 mL high purity water)

Synonym(s):

Urea solution, Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.25

form

powder

concentration

8 M (after reconstitution with 16 mL high purity water)

solubility

H2O: soluble (1 vial plus 16 mL)

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Urea is a chaotropic agent and is used for protein denaturation. It disturbs the hydrogen bonds in the secondary, tertiary and quaternary structure of proteins. It can also disturb hydrogen bonding present in DNA secondary structure.

Application

Useful for solubilizing and denaturing proteins in preparation for 2D-PAGE, enzymatic digests, IMAC Chromatography, and related procedures.
Urea has been used for protein sample preparation for reverse-phase nanospray liquid chromatography-tandem mass spectrometry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheehan D
Physical Biochemistry: Principles and Applications (2009)
MPYS, a novel membrane tetraspanner, is associated with major histocompatibility complex class II and mediates transduction of apoptotic signals.
Jin L, et al.
Molecular and Cellular Biology, 28, 5014-5026 (2008)
L Li et al.
Cell death and differentiation, 22(7), 1081-1093 (2014-12-20)
P53 is critically important in preventing oncogenesis but its role in inflammation in general and in the function of inflammatory macrophages in particular is not clear. Here, we show that bone marrow-derived macrophages exhibit endogenous p53 activity, which is increased
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
Marco Benevento et al.
Nature communications, 5, 5613-5613 (2014-12-11)
The ectopic expression of Oct4, Klf4, c-Myc and Sox2 (OKMS) transcription factors allows reprogramming of somatic cells into induced pluripotent stem cells (iPSCs). The reprogramming process, which involves a complex network of molecular events, is not yet fully characterized. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service