Skip to Content
Merck
All Photos(2)

Documents

1.02950

Supelco

Dimethyl sulfoxide

for spectroscopy Uvasol®

Synonym(s):

Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-664-3
NACRES:
NA.03

vapor density

2.7 (vs air)

Quality Level

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

573 °F

potency

28300 mg/kg LD50, oral (Rat)
40000 mg/kg LD50, skin (Rat)

expl. lim.

42 %, 63 °F

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤0.0004%

color

APHA: ≤10

transmittance

270 nm, ≥35%
280 nm, ≥50%
310 nm, ≥80%
330 nm, ≥90%
350 nm, ≥97%

refractive index

n20/D 1.479 (lit.)

kinematic viscosity

2.14 cSt

bp

189 °C (lit.)

mp

16-19 °C (lit.)

transition temp

flash point 87 °C

density

1.10 g/mL (lit.)

UV absorption

λ: 270 nm Amax: ≤0.46
λ: 280 nm Amax: ≤0.30
λ: 310 nm Amax: ≤0.10
λ: 330 nm Amax: ≤0.05
λ: 350 nm Amax: ≤0.013

storage temp.

2-30°C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Specifications

Saturation Concentration in Air: 8.0 g/m3 (20 °C)

Analysis Note

Purity (GC): ≥ 99.8 %
Evaporation residue: ≤ 0.0004 %
Water: ≤ 0.05 %
Color: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 270 nm): ≥ 35 %
Transmission (at 280 nm): ≥ 50 %
Transmission (at 310 nm): ≥ 80 %
Transmission (at 330 nm): ≥ 90 %
Transmission (from 350 nm): ≥ 97 %
Absorbance (at 270 nm): ≤ 0.46
Absorbance (at 280 nm): ≤ 0.30
Absorbance (at 310 nm): ≤ 0.10
Absorbance (at 330 nm): ≤ 0.05
Absorbance (from 350 nm): ≤ 0.013

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service