Skip to Content
Merck
All Photos(2)

Key Documents

1.00765

Supelco

Boric acid

for inorganic trace analysis, 99.9999 Suprapur®

Synonym(s):

Orthoboric acid, Trihydroxidoboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
MDL number:
UNSPSC Code:
12352301
EC Index Number:
233-139-2
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

vapor pressure

2.6 mmHg ( 20 °C)

Assay

≥99% (acidimetric)
99.9999%

form

solid

potency

2660 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

3.8-4.8 (20 °C, 33 g/L in H2O)

mp

160 °C (dec.) (lit.)

solubility

49.2 g/L

density

1.489 g/cm3 at 23 °C

bulk density

400‑600 kg/m3

anion traces

chloride (Cl-): ≤3 ppm
phosphate (PO43-): ≤0.5 ppm
silicate (as SiO2): ≤0.5 ppm
sulfate (SO42-): ≤5 ppm

cation traces

Ag: ≤0.010 ppm
Al: ≤0.050 ppm
As: ≤0.050 ppm
Au: ≤0.020 ppm
Ba: ≤0.050 ppm
Be: ≤0.010 ppm
Ca: ≤0.050 ppm
Cd: ≤0.010 ppm
Co: ≤0.010 ppm
Cr: ≤0.010 ppm
Cu: ≤0.010 ppm
Fe: ≤0.050 ppm
Ga: ≤0.050 ppm
In: ≤0.020 ppm
K: ≤0.10 ppm
Li: ≤0.010 ppm
Mg: ≤0.10 ppm
Mn: ≤0.010 ppm
Mo: ≤0.010 ppm
N: ≤0.010 ppm
Na: ≤0.050 ppm
Pb: ≤0.020 ppm
Sr: ≤0.010 ppm
Tl: ≤0.020 ppm
V: ≤0.010 ppm
Zn: ≤0.010 ppm

SMILES string

OB(O)O

storage temp.

2-30°C

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Systematic development of immunohistochemistry protocol for large cryosections-specific to non-perfused fetal brain.: This study by Pandurangan et al. highlights the role of boric acid in developing an optimized immunohistochemistry protocol for large cryosections of non-perfused fetal brain tissues. Boric acid is used as a buffer component to enhance antigen retrieval and staining specificity, showcasing its importance in neuroanatomical research (Pandurangan et al., 2024).

  • Proteomic Analysis of Rat Duodenum Reveals the Modulatory Effect of Boron Supplementation on Immune Activity.: Zhao et al. investigate the impact of boric acid on immune activity through proteomic analysis of rat duodenum. The study demonstrates boric acids role in modulating protein expression related to immune responses, providing insights into its potential applications in immunology and nutritional biochemistry (Zhao et al., 2023).

  • The impact of boron nutrient supply in mulberry (Morus alba) response to metabolomics, enzyme activities, and physiological parameters.: Zhang et al. explore the effects of boric acid on mulberry plants′ metabolic profiles, enzyme activities, and physiological parameters. This research underscores boric acids significance in plant nutrition and stress response, offering valuable data for agricultural chemistry and plant sciences (Zhang et al., 2023).

Analysis Note

Purity (metallic): ≥ 99.9999 %
Assay (acidimetric): ≥ 99.0 %
Chloride (Cl): ≤ 3 ppm
Phosphate (PO₄): ≤ 0.5 ppm
Silicate (as SiO₂): ≤ 0.5 ppm
Sulfate (SO₄): ≤ 5 ppm
Ag (Silver): ≤ 0.010 ppm
Al (Aluminium): ≤ 0.050 ppm
As (Arsenic): ≤ 0.050 ppm
Au (Gold): ≤ 0.020 ppm
Ba (Barium): ≤ 0.050 ppm
Be (Beryllium): ≤ 0.010 ppm
Ca (Calcium): ≤ 0.050 ppm
Cd (Cadmium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.010 ppm
Fe (Iron): ≤ 0.050 ppm
Ga (Gallium): ≤ 0.050 ppm
In (Indium): ≤ 0.020 ppm
K (Potassium): ≤ 0.10 ppm
Li (Lithium): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Mo (Molybdenum): ≤ 0.010 ppm
Na (Sodium): ≤ 0.050 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.020 ppm
Sr (Strontium): ≤ 0.010 ppm
Tl (Thallium): ≤ 0.020 ppm
V (Vanadium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service