Skip to Content
Merck
All Photos(1)

Documents

07-140

Sigma-Aldrich

Anti-STAT2 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-IMD44, Anti-ISGF-3, Anti-P113, Anti-PTORCH3, Anti-STAT113

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... STAT2(6773)

Specificity

STAT2

Immunogen

GST-fusion protein corresponding to residues 672-925 of murine STAT2

Application

Anti-STAT2 Antibody is a Rabbit Polyclonal Antibody for detection of STAT2 also known as Signal Transducer & Activator of Transcription 2 & has been tested in EMSA, IP, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on

Target description

130kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Affinity Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Matsumoto et al.
Biological chemistry, 380(6), 699-703 (1999-08-03)
The interferon-stimulated gene factor 3 (ISGF3) transcription factor has been extensively studied in the context of the type I interferon (IFN-alpha/beta)-mediated antiviral response; it consists of the major DNA-binding component p48, and the signal transducers and activators of transcription (Stat)1
Genetic manipulation of the ApoF/Stat2 locus supports an important role for type I interferon signaling in atherosclerosis.
Lagor, WR; Fields, DW; Bauer, RC; Crawford, A; Abt, MC; Artis, D; Wherry, EJ; Rader, DJ
Atherosclerosis null
A distal regulatory region is required for constitutive and IFN-beta-induced expression of murine TLR9 gene.
Zhu Guo, Sanjay Garg, Karen M Hill, Lakshmi Jayashankar, Myesha R Mooney, Mary Hoelscher et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Connie C Qiu et al.
Oncoimmunology, 10(1), 1860477-1860477 (2021-01-19)
STAT2 is a central component of the ISGF3 transcriptional complex downstream of type I interferon (IFN-I) signaling. The significance of in vivo IFN-I/STAT1 signals in cDCs is well-established in the generation of antitumor cytotoxic T cell (CTL) responses. However, the role
Reovirus mu2 protein inhibits interferon signaling through a novel mechanism involving nuclear accumulation of interferon regulatory factor 9.
Zurney, J; Kobayashi, T; Holm, GH; Dermody, TS; Sherry, B
Journal of virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service