Skip to Content
Merck
All Photos(3)

Key Documents

K2144

Sigma-Aldrich

Ko143 hydrate

≥98% (HPLC), powder, BCRP inhibitor

Synonym(s):

(3S,6S,12aS)-1,2,3,4,6,7,12,12a-Octahydro-9-methoxy-6-( 2-methylpropyl)-1,4-dioxopyrazino[1′,2′:1,6]pyrido[3,4- b]indole-3-propanoic acid 1,1-dimethylethyl ester hydrate, Ko-143 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35N3O5 · xH2O
CAS Number:
Molecular Weight:
469.57 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Ko143 hydrate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

room temp

SMILES string

O.COc1ccc2c3C[C@@H]4N([C@@H](CC(C)C)c3[nH]c2c1)C(=O)[C@H](CCC(=O)OC(C)(C)C)NC4=O

InChI

1S/C26H35N3O5.H2O/c1-14(2)11-20-23-17(16-8-7-15(33-6)12-19(16)27-23)13-21-24(31)28-18(25(32)29(20)21)9-10-22(30)34-26(3,4)5;/h7-8,12,14,18,20-21,27H,9-11,13H2,1-6H3,(H,28,31);1H2/t18-,20-,21-;/m0./s1

InChI key

ZXXZDXPNJJZRDC-PSLBYKFTSA-N

Application

Ko143 hydrate has been used:
  • to determine the role of ATP-binding cassette sub-family G member 2 (ABCG2), human embryonic kidney (HEK)-C1 and HEK-ABCG2 in tumor microenvironment
  • to inhibit ABCG2 for sphere formation assay
  • in calcein-AM efflux inhibition to monitor multidrug resistance protein (MRP)-function in kidney
  • for cell viability assay

Biochem/physiol Actions

Ko143 has been used as a positive control inhibitor on functions of BCRP in MCF7 and BCRP over-expressing MCF7/MX100 cell lines using a BCRP prototypical substrate mitoxantrone.
Ko143 has been used as a positive control inhibitor on functions of BCRP in MCF7 and BCRP over-expressing MCF7/MX100 cell lines using a BCRP prototypical substrate mitoxantrone. The ABCG2 transporter breast cancer resistance protein (BCRP) plays an important role in disposition of many drugs and environmental toxins. Ko143 displays > 200-fold selectivity over P-gp and MRP-1 transporters. It increases intracellular drug accumulation and reverse BCRP-mediated multidrug resistance. It blocked topotecan and ABZSO transport in a concentration-dependent manner. Ko143 is reported to be a more specific inhibitor of BCRP than other known inhibitors of BCRP such as fumitremorgin C and GF120918.
Ko143 has been used as a positive control inhibitor on functions of breast cancer resistance protein (BCRP) using a BCRP prototypical substrate mitoxantrone.
Ko143 has been used as a positive control inhibitor on functions of breast cancer resistance protein (BCRP) using a BCRP prototypical substrate mitoxantrone. BCRP, an ABCG2 transporter, plays an important role in disposition of many drugs and environmental toxins. Ko143 displays > 200-fold selectivity over P-gp and MRP-1 transporters and thus is more specific than other known BCRP inhibitors such as fumitremorgin C and GF120918. It increases intracellular drug accumulation and reverses BCRP-mediated multidrug resistance. It blocks topotecan and ABZSO transport in a concentration-dependent manner.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage and Stability

The stability of this product in solution has not been determined. According to various sources this product can be stored in solvent (DMSO) at -80°C for 6 months and -20°C for 1 month.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Architecture of chimeric spheroids controls drug transport
Curran S, et al.
Cancer Microenvironment, 8(2), 101-109 (2015)
Multi-drug resistance ABC transporter inhibition enhances murine ventral prostate stem/progenitor cell differentiation
Samant MD, et al.
Stem Cells and Development, 24(10), 1236-1251 (2015)
Overcoming resistance to the THZ series of covalent transcriptional CDK inhibitors
Gao Y, et al.
Cell Chemical Biology, 25(2), 135-142 (2018)
Lu Xiao et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 93, 456-467 (2016-09-03)
Efflux transporters are extensively distributed and expressed in the intestinal epithelium and contribute to the low oral bioavailability of flavonoids and flavonoid glucuronides by pumping these compounds back into intestinal lumen. Our previous study has shown the inhibitory effect of
Cong Duan et al.
Biopharmaceutics & drug disposition, 38(1), 75-83 (2016-12-08)
Lindera aggregata (Sims) Kosterm root has been used in traditional Chinese medicine for the treatment of rheumatism palsy, dyspepsia and frequent urination for a long time. Norisoboldine, the main active constituent of this herb drug, possesses outstanding anti-arthritis activity. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service