Skip to Content
Merck
All Photos(1)

Key Documents

05-824

Sigma-Aldrich

Anti-Estrogen Receptor β Antibody, clone 68-4, rabbit monoclonal

clone 68-4, Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

68-4, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ESR2(2100)

General description

Estrogen receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of ligand-activated transcription factors. Estrogen receptors, including ERα and ERβ, contain DNA binding and ligand binding domains and are critically involved in regulating the normal function of reproductive tissues. They are located in the nucleus, though some estrogen receptors associate with the cell surface membrane and can be rapidly activated by exposure of cells to estrogen. ERα and ERβ have been shown to be differentially activated by various ligands. Receptor-ligand interactions trigger a cascade of events, including dissociation from heat shock proteins, receptor dimerization, phosphorylation and the association of the hormone activated receptor with specific regulatory elements in target genes. Evidence suggests that ERα and ERβ may be regulated by distinct mechanisms even though they share many functional characteristics.
Produced in collaboration with Epitomics

Specificity

Predicted to cross-react with rat and mouse based on sequence homology
Recognizes Estrogen Receptor β, long form, does not detect estrogen receptor alpha.

Immunogen

KLH-conjugated linear peptide corresponding to amino acids 108-127 from the N-terminal half of human estrogen receptor beta.

Application

Detect Estrogen Receptor β also known as Nuclear receptor subfamily 3 group A member 2 with Anti-Estrogen Receptor β Antibody, clone 68-4 (Rabbit Monoclonal Antibody), that has been demonstrated to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on RIPA lysates from MCF7 cells

Target description

59 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% sodium azide, pH 7.4.
Protein A purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF7 cell extracts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robynn V Schillace et al.
BMC cancer, 14, 733-733 (2014-10-02)
Estrogen receptor positive breast cancers have high recurrence rates despite tamoxifen therapy. Breast cancer stem/progenitor cells (BCSCs) initiate tumors, but expression of estrogen (ER) or progesterone receptors (PR) and response to tamoxifen is unknown. Interleukin-6 (IL-6) and interleukin-8 (IL-8) may
V Praveen Chakravarthi et al.
Endocrinology, 161(4) (2020-03-07)
Over the entire reproductive lifespan in mammals, a fixed number of primordial follicles serve as the source of mature oocytes. Uncontrolled and excessive activation of primordial follicles can lead to depletion of the ovarian reserve. We observed that disruption of
Up-regulation of estrogen receptor-beta expression during osteogenic differentiation of human periodontal ligament cells.
X Tang, H Meng, J Han, L Zhang, J Hou, F Zhang
Journal of Periodontal Research null
M A Karim Rumi et al.
Endocrinology, 158(7), 2330-2343 (2017-05-19)
Estrogens are essential hormones for the regulation of fertility. Cellular responses to estrogens are mediated by estrogen receptor α (ESR1) and estrogen receptor β (ESR2). In mouse and rat models, disruption of Esr1 causes infertility in both males and females.
Yan Zhu et al.
Science (New York, N.Y.), 295(5554), 505-508 (2002-01-19)
Blood vessels express estrogen receptors, but their role in cardiovascular physiology is not well understood. We show that vascular smooth muscle cells and blood vessels from estrogen receptor beta (ERbeta)-deficient mice exhibit multiple functional abnormalities. In wild-type mouse blood vessels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service