Skip to Content
Merck
All Photos(1)

Key Documents

05-592

Sigma-Aldrich

Anti-PP2A Antibody, B subunit, clone 2G9

ascites fluid, clone 2G9, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2G9, monoclonal

species reactivity

rat, bovine, mouse, Xenopus, rabbit, pig, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... PPP2CA(5515)

Specificity

PP2A B subunit, isoform reactivity is α > δ,β > γ

Immunogen

Peptide corresponding to residues 398-411 of human PP2A, B subunit

Application

Detect PP2A using this Anti-PP2A Antibody, B subunit, clone 2G9 validated for use in IH, IP & WB.
Research Category
Signaling

Neuroscience
Research Sub Category
Kinases & Phosphatases

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on RIPA lysates from non-stimulated A431 cells or mouse 3T3/A31 cells

Target description

55kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and glycerol to 30%
Ascites

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilker K Sariyer et al.
Virology, 375(2), 464-479 (2008-03-21)
Previous studies have demonstrated that the JC virus (JCV) late regulatory protein agnoprotein is phosphorylated by the serine/threonine-specific protein kinase-C (PKC) and mutants of this protein at the PKC phosphorylation sites exhibit defects in the viral replication cycle. We have
Jennifer B Jackson et al.
Neoplasia (New York, N.Y.), 14(7), 585-599 (2012-08-21)
Heterotrimeric protein phosphatase 2A (PP2A) consists of catalytic C (PP2Ac), structural A, and regulatory B-type subunits, and its dysfunction has been linked to cancer. Reversible methylation of PP2Ac by leucine carboxyl methyltransferase 1 (LCMT-1) and protein phosphatase methylesterase 1 (PME-1)
Protein phosphatase 2A regulates estrogen receptor alpha (ER) expression through modulation of ER mRNA stability.
Keen, JC; Zhou, Q; Park, BH; Pettit, C; Mack, KM; Blair, B; Brenner, K; Davidson, NE
The Journal of Biological Chemistry null
Chian Ju Jong et al.
The Journal of biological chemistry, 295(17), 5654-5668 (2020-03-12)
Protein phosphatase 2A (PP2A) is a large enzyme family responsible for most cellular Ser/Thr dephosphorylation events. PP2A substrate specificity, localization, and regulation by second messengers rely on more than a dozen regulatory subunits (including B/R2, B'/R5, and B″/R3), which form
Jocelyn A Lee et al.
The Journal of biological chemistry, 293(25), 9636-9650 (2018-05-08)
Leucine carboxyl methyltransferase-1 (LCMT-1) methylates the C-terminal leucine α-carboxyl group of the catalytic subunits of the protein phosphatase 2A (PP2A) subfamily of protein phosphatases, PP2Ac, PP4c, and PP6c. LCMT-1 differentially regulates the formation and function of a subset of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service