Skip to Content
Merck
All Photos(1)

Key Documents

04-1570

Sigma-Aldrich

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12

clone 4E12, from rat

Synonym(s):

DNA-directed RNA polymerase II A, DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA polymerase II subunit B1, RNA-directed RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E12, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

ChIP: suitable
ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer7)

Gene Information

human ... POLR2B(5431)

General description

RNA polymerase II subunit B1 (RPB1) is the largest subunit of the RNA polymerase II complex. As a holoenzyme RNA polymerase II catalyzes transcription of eukaryotic DNA into RNA using the four ribonucleoside triphosphates as substrates. The RBP1 subunit, in combination with other polymerase subunits, forms a large central cleft that maintains contact between the active site of the enzyme, the DNA template, and the nascent RNA transcript. This subunit also contains a carboxy terminal domain (CTD) consisting of tandem heptapeptide repeats. Phosphorylation activates the RNA polymerase II beta subunit, allowing it to serve as an assembly platform for additional subunits that modulate initiation, elongation, termination and mRNA processing. In actively transcribing RNA polymerase ‘Ser-2’ and ‘Ser-5’ of the heptapeptide repeat are phosphorylated. Ser-7 is phosphorylated before initiation of transcription at promoter regions.

Specificity

This antibody recognizes RNA polymerase II subunit B1 at the CTD when phosphorylated at Ser7.

Immunogen

Epitope: Ser7
Ovalbumin-conjugated linear peptide corrresponding to human RNA polymerase subunit B1 CTD phosphorylated at Ser7.

Application

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12 is a Rat monoclonal antibody for detection of RNA polymerase II subunit B1 (phospho-CTD Ser-7) has been validated in WB, ELISA.
Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in ChIP. (Chapman, R., et al. (2007). Science. 318(5857):1780 -1782.)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in γ-PPase untreated and treated NIH/3T3 cell lysates.

Western Blot Analysis: 0.25 µg/ml of this antibody detected RNA polymerase II CTD on 10 µg of γ-PPase untreated and treated NIH/3T3 cell lysates.

Target description

~ 220 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
γ-protein phosphatase (γ-Ppase) untreated and treated NIH/3T3 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana M Sanchez et al.
Nucleic acids research, 48(9), 4811-4826 (2020-04-14)
The phosphorylation pattern of Pol2 CTD Y1S2P3T4S5P6S7 repeats comprises an informational code coordinating transcription and RNA processing. cis-trans isomerization of CTD prolines expands the scope of the code in ways that are not well understood. Here we address this issue
Tianlu Huang et al.
Cell death & disease, 10(8), 602-602 (2019-08-11)
Cholangiocarcinoma (CCA) is a fatal disease without effective targeted therapy. We screened a small-molecule library of 116 inhibitors targeting different targets of the cell cycle and discovered several kinases, including Cyclin-dependent kinase 7 (CDK7) as vulnerabilities in CCA. Analysis of
Chao Han et al.
PloS one, 15(11), e0240801-e0240801 (2020-11-03)
Our previously study shown that Lysophosphatidylcholine Acyltransferase1 (LPCAT1) is overexpressed in castration resistant prostate cancer (CRPC) relative to primary prostate cancer (PCa), and androgen controls its expression via the Wnt signaling pathway. While highly expressed in CRPC, the role of
Gregory T Booth et al.
Nature communications, 9(1), 543-543 (2018-02-09)
Post-translational modifications of the transcription elongation complex provide mechanisms to fine-tune gene expression, yet their specific impacts on RNA polymerase II regulation remain difficult to ascertain. Here, in Schizosaccharomyces pombe, we examine the role of Cdk9, and related Mcs6/Cdk7 and
Emily Brookes et al.
Cell stem cell, 10(2), 157-170 (2012-02-07)
Polycomb repressor complexes (PRCs) are important chromatin modifiers fundamentally implicated in pluripotency and cancer. Polycomb silencing in embryonic stem cells (ESCs) can be accompanied by active chromatin and primed RNA polymerase II (RNAPII), but the relationship between PRCs and RNAPII

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service