Skip to Content
Merck
All Photos(1)

Key Documents

T-035

Supelco

O-Desmethyl-cis-tramadol hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24ClNO2
CAS Number:
Molecular Weight:
285.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@]1([C@@H](CN(C)C)CCCC1)C2=CC=CC(O)=C2.Cl

InChI

1S/C15H23NO2.ClH/c1-16(2)11-13-6-3-4-9-15(13,18)12-7-5-8-14(17)10-12;/h5,7-8,10,13,17-18H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1

InChI key

IRGWVAWLHXDKIX-PBCQUBLHSA-N

General description

O-Desmethyltramadol is an opioid analgesic and an active primary blood metabolite of the synthetic analgesic tramadol. O-Desmethyltramadol is the active component in the designer drug “Krypton”, which is marketed in many countries as a legal high. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as urine drug testing, pain prescription monitoring, forensic analysis, and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service