Skip to Content
Merck
All Photos(1)

Key Documents

N-053

Supelco

Norhydrocodone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Norhydrocodone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3·HCl
CAS Number:
Molecular Weight:
321.80
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.COc1ccc2C[C@H]3NCC[C@@]45[C@H]3CCC(=O)[C@@H]4Oc1c25

InChI

1S/C17H19NO3.ClH/c1-20-13-5-2-9-8-11-10-3-4-12(19)16-17(10,6-7-18-11)14(9)15(13)21-16;/h2,5,10-11,16,18H,3-4,6-8H2,1H3;1H/t10-,11+,16-,17-;/m0./s1

InChI key

IQVZQPJFFRBEHK-NRGUFEMZSA-N

General description

This Certified Spiking Solution® is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications including pain prescription monitoring, urine drug testing, forensic analysis, clinical toxicology, and pharmaceutical research. Norhydrocodone is an inactive primary urinary metabolite of the analgesic hydrocodone. Hydrocodone, a semi-synthetic opioid derived from codeine and thebaine, is used as a painkiller and as an antitussive.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service