Skip to Content
Merck
All Photos(1)

Key Documents

C-174

Supelco

4-Chloromethcathinone (4-CMC HCl) hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClNO · HCl
Molecular Weight:
234.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)C(=O)C(NC)C.Cl

InChI

1S/C10H12ClNO.ClH/c1-7(12-2)10(13)8-3-5-9(11)6-4-8;/h3-7,12H,1-2H3;1H

InChI key

WJAQVQFSQUVTAY-UHFFFAOYSA-N

General description

4-Chloromethcathinone, also known as 4-CMC, is a stimulant and designer drug found in "bath salts" sold at head shops and over the Internet. These synthetic cathinones offer recreational uses that mimic the effects of illegal drugs such as cocaine, methamphetamine, and LSD. This certified solution standard is suitable for numerous GC/MS and LC/MS applications including clinical toxicology testing, forensic analysis, urine drug testing or calibrator preparation.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meltem Weger et al.
Physiology & behavior, 186, 31-36 (2018-01-15)
Social rank is frequently established through aggressive encounters between new conspecifics. Despite increasing evidence suggesting that social rank is critical for the well-being of both humans and animals, knowledge about the factors influencing social rank remain scarce. Stress was previously
Maaike Schilperoort et al.
Aging cell, 20(10), e13474-e13474 (2021-10-01)
Glucocorticoid (GC)-induced osteoporosis is a widespread health problem that is accompanied with increased fracture risk. Detrimental effects of anti-inflammatory GC therapy on bone have been ascribed to the excess in GC exposure, but it is unknown whether there is also
C Bouarab et al.
Molecular psychiatry, 26(9), 4968-4981 (2021-01-30)
Moderate stress increases memory and facilitates adaptation. In contrast, intense stress can induce pathological memories as observed in post-traumatic stress disorders (PTSD). A shift in the balance between the expression of tPA and PAI-1 proteins is responsible for this transition.
Wei-Li Wu et al.
Nature, 595(7867), 409-414 (2021-07-02)
Social interactions among animals mediate essential behaviours, including mating, nurturing, and defence1,2. The gut microbiota contribute to social activity in mice3,4, but the gut-brain connections that regulate this complex behaviour and its underlying neural basis are unclear5,6. Here we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service