Skip to Content
Merck
All Photos(1)

Key Documents

H-101

Supelco

trans-3′-Hydroxycotinine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1[C@H](C2=CN=CC=C2)C[C@@H](O)C1=O

InChI

1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9+/m0/s1

InChI key

XOKCJXZZNAUIQN-DTWKUNHWSA-N

General description

Trans-3′-Hydroxycotinine is a major nicotine metabolite excreted in the urine of smokers and users of other tobacco or nicotine products including cigars, chewing tobacco and snuff. This Certified Spiking Solution® is suitable for use in nicotine testing methods or clinical and diagnostic testing of nicotine biomarkers by LC-MS/MS or GC/MS.

Application


  • Wastewater-Based Tobacco Use Estimates: trans-3′-Hydroxycotinine is used as WBE biomarkers to to estimate tobacco use. It has significantly improved wastewater-based tobacco use estimates. This application enhances our understanding of public health trends by providing more accurate measurements of tobacco consumption in populations, using advanced environmental monitoring techniques (Zheng et al., 2023).

  • Predicting Nicotine Metabolism: Trans-3′-Hydroxycotinine is utilized in genomic studies to predict nicotine metabolism across different ancestries, which helps in tailoring smoking cessation therapies based on individual genetic profiles.(Baurley et al., 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meilan Yu et al.
Biotechnology letters, 37(8), 1615-1622 (2015-05-16)
The action modes of an endocellulase, EGA, and its domains (CD9 and CBM3) during enzymatic treatment of cotton fabrics were investigated. EGA, CD9 and CBM3 had the binding capacity to cellulose substrates, of which the filter paper was the substrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service