Skip to Content
Merck
All Photos(1)

Key Documents

C-016

Supelco

(−)-Cotinine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1C(CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1

InChI key

UIKROCXWUNQSPJ-VIFPVBQESA-N

General description

An analytical reference standard of the major urinary metabolite of nicotine. Not only is cotinine a biomarker for users of nicotine products including cigarettes, cigars, and snuff, but it′s also an alkaloid found in tobacco. Suitable uses for this analytical reference standard include as a starting material in calibrators and controls for GC/MS and LC/MS applications in nicotine testing methods or clinical and diagnostic testing of nicotine biomarkers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Haufroid et al.
International archives of occupational and environmental health, 71(3), 162-168 (1998-05-20)
A minireview is presented concerning the use of cotinine as a tobacco-smoke exposure index. First, general considerations about methods for the determination of urinary cotinine are presented. Besides pure analytical aspects, this minireview considers major problems encountered in the establishment
Kuan-Yen Tung et al.
Environmental research, 124, 7-12 (2013-04-30)
Environmental tobacco smoke (ETS) is a hazardous component of indoor air, and may increase the risk of respiratory diseases, atherosclerosis and otitis media in children. In this study, we explored the relationship between time inside the house, ETS exposure and
Ines Moreno-Gonzalez et al.
Nature communications, 4, 1495-1495 (2013-02-21)
Several epidemiological studies have shown that cigarette smoking might alter the incidence of Alzheimer's disease. However, inconsistent results have been reported regarding the risk of Alzheimer's disease among smokers. Previous studies in experimental animal models have reported that administration of
Cotinine as a biomarker of environmental tobacco smoke exposure.
N L Benowitz
Epidemiologic reviews, 18(2), 188-204 (1996-01-01)
Brian K Kit et al.
Pediatrics, 131(3), 407-414 (2013-02-13)
To examine exposure to tobacco smoke products (TSPs), environmental tobacco smoke (ETS), and in-home smoke among youth with asthma in the United States. Nationally representative, cross-sectional data from 2250 youth aged 4 to 19 years with current asthma in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service